Skip to Content
Merck
All Photos(1)

Key Documents

MABE281

Sigma-Aldrich

Anti-MDM2 Antibody, clone 2A10

clone 2A10, from mouse

Synonym(s):

E3 ubiquitin-protein ligase Mdm2, Double minute 2 protein, Hdm2, Oncoprotein Mdm2, p53-binding protein Mdm2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

2A10, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MDM2(4193)

General description

MDM2 is an E3 ubiquitin ligase that is most widely studied for its role in regulating the p53 tumor suppressor. MDM2 antagonizes the transcriptional activity of p53 in two main ways. The N-terminal of MDM2 interacts and disables the transactivation domain of p53. In addition, the C-terminal E3 ligase region of MDM2 can attach monoubiquitin to p53 resulting in the transport of p53 from the nucleus to the cytoplasm, thereby preventing the interaction of p53 with target DNA sequences. In addition, MDM2 may also attach polyubiquitin chains to p53, resulting in the degradation of p53 by the proteosome. MDM2 may produce these effects in cooperation with the Mdmx protein. Overall, MDM2 inhibits the ability of p53 to induce cell cycle arrest or apoptosis, and may contribute to oncogenesis.

Application

Anti-MDM2 Antibody, clone 2A10 is a Mouse Monoclonal Antibody for detection of MDM2 also known as E3 ubiquitin-protein ligase Mdm2 or Hdm2 & has been validated in WB & IP.
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in human lymphoblastoid cell line (NL553) treated with NCS. (Khosravi, R., et al. (1999). PNAS. 96(26):14973–14977.)

Quality

Evaluated by Western Blot in A-549 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected MDM2 on 10 µg of A-549 cell lysate.

Target description

~90 kDa observed. The calculated molecular weight is 55 kDa, however MDM2 has been shown as a ~90 kDa band in western blots (Erhardt, P., et al. (1997). The Journal of Biological Chemistry. 272:15049-15052.). Uniprot describes many isoforms between 12-56 kDa, which can be highly phosphorylated and ubiquitinated.

Physical form

Format: Purified
Purified mouse monoclonal IgG2aκ cultured supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Analysis Note

Control
A-549 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vinod Pant et al.
Genes & development, 27(17), 1857-1867 (2013-08-27)
The p53-Mdm2 feedback loop is perceived to be critical for regulating stress-induced p53 activity and levels. However, this has never been tested in vivo. Using a genetically engineered mouse with mutated p53 response elements in the Mdm2 P2 promoter, we
Shunbin Xiong et al.
Cancer research, 70(18), 7148-7154 (2010-08-26)
High levels of the critical p53 inhibitor Mdm4 is common in tumors that retain a wild-type p53 allele, suggesting that Mdm4 overexpression is an important mechanism for p53 inactivation during tumorigenesis. To test this hypothesis in vivo, we generated transgenic
Oliver Bischof et al.
Molecular cell, 22(6), 783-794 (2006-06-24)
Cellular senescence and apoptosis have evolved to restrain unwarranted proliferation of potentially tumorigenic cells. Here we show that overexpression of the E3 SUMO ligase PIASy in normal human fibroblasts recruits the p53 and Rb tumor suppressor pathways to provoke a
Kyle P Feeley et al.
Cancer research, 77(14), 3823-3833 (2017-06-04)
p53 deletion prevents the embryonic lethality of normal tissues lacking Mdm2, suggesting that cells can survive without Mdm2 if p53 is also absent. Here we report evidence challenging this view, with implications for therapeutically targeting Mdm2. Deletion of Mdm2 in
Andrés Pizzorno et al.
Scientific reports, 8(1), 3746-3746 (2018-03-01)
The interplay between influenza A viruses (IAV) and the p53 pathway has been reported in several studies, highlighting the antiviral contribution of p53. Here, we investigated the impact of IAV on the E3-ubiquitin ligase Mdm2, a major regulator of p53

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service