Skip to Content
Merck
All Photos(1)

Key Documents

MAB837

Sigma-Aldrich

Anti-Papillomavirus Antibody, 1, 6, 11, 16, 18, 31, clone 1H8

clone 1H8, Chemicon®, from mouse

Synonym(s):

HPV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

1H8, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

Specificity

Human papillomavirus proteins - HPV-1, HPV-6, HPV-11, HPV-16, HPV-18 and HPV-31.

Immunogen

Epitope: 1, 6, 11, 16, 18, 31
SDS-disrupted bovine papillomavirus type 1.

Application

Anti-Papillomavirus Antibody, 1, 6, 11, 16, 18, 31, clone 1H8 is an antibody against Papillomavirus for use in ELISA, WB, IH.
Immunohistochemistry

ELISA

Western Blot

Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Liquid in 0.02M PB 0.25M NaCl, and 0.1% sodium azide, pH 7.6.

Storage and Stability

Maintain at 2-8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determinants of autoantibody induction by conjugated papillomavirus virus-like particles.
Bryce Chackerian, Petra Lenz, Douglas R Lowy, John T Schiller
Journal of immunology (Baltimore, Md. : 1950) (1950)
Anna Yemelyanova et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 26(2), 268-274 (2012-09-22)
While cervical cancer screening relies on cervical cytology and high-risk human papillomavirus (HPV) detection, the histologic diagnosis, and specifically lesion grade, is the main parameter that drives clinical management of screen-positive women. Morphologically diagnosed squamous intraepithelial lesions (SIL/CIN) regress spontaneously
Neutralization of bovine papillomavirus by antibodies to L1 and L2 capsid proteins
Roden, R B, et al
Journal of virology, 68, 7570-7574 (1994)
A A McBride et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(10), 5534-5539 (2000-05-11)
Bovine papillomavirus type 1 (BPV-1) induces fibropapillomas in its natural host and can transform fibroblasts in culture. The viral genome is maintained as an episome within fibroblasts, which has allowed extensive genetic analyses of the viral functions required for DNA
Identification of the bovine papillomavirus L1 gene product using monoclonal antibodies.
Cowsert, L M, et al.
Virology, 165, 613-615 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service