Skip to Content
Merck
All Photos(1)

Key Documents

MAB8261

Sigma-Aldrich

Anti-Influenza A Antibody, H1N1, clone 9B3.2

clone 9B3.2, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

9B3.2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Influenza A H1N1 antigen. No reactivity shown to Influenza B strains.

SPECIES REACTIVITY:

Reacts strongly with the following H1N1 strain: Beijing. A/ Texas /36/91, A/Berkeley/1/98, A/HongKong/503/97,A/Nanchang /16A/98,A/PR8/34 and New Caledoniastrain.

Immunogen

Epitope: H1N1
Influenza blend

Application

IF

Optimal dilutions must be determined by end user
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Influenza A Antibody, H1N1, clone 9B3.2 is validated for use in IF for the detection of Influenza A.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02 M Phosphate Buffer + 0.25 M Sodium Chloride + 0.1% Sodium Azide, pH = 7.6

Storage and Stability

Store at 2° to 8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sialylneolacto-N-tetraose c (LSTc)-bearing liposomal decoys capture influenza A virus.
Hendricks, GL; Weirich, KL; Viswanathan, K; Li, J; Shriver, ZH; Ashour, J; Ploegh et al.
The Journal of Biological Chemistry null
Claudia Bank et al.
Evolution; international journal of organic evolution, 70(11), 2470-2484 (2016-11-01)
The rapid evolution of drug resistance remains a critical public health concern. The treatment of influenza A virus (IAV) has proven particularly challenging, due to the ability of the virus to develop resistance against current antivirals and vaccines. Here, we
Michal Mendelson et al.
Journal of virology, 84(8), 3789-3797 (2010-02-12)
Natural killer (NK) cells serve as a crucial first-line defense against tumors and virus-infected cells. We previously showed that lysis of influenza virus (IV)-infected cells is mediated by the interaction between the NK receptor, NKp46, and the IV hemagglutinin (HA)
Jeannine A Majde et al.
Journal of neurovirology, 13(5), 399-409 (2007-11-13)
Influenza pneumonitis causes severe systemic symptoms in mice, including hypothermia and excess sleep. The association of extrapulmonary virus, particularly virus in the brain, with the onset of such disease symptoms has not been investigated. Mature C57BL/6 male mice were infected
Kristina L Prachanronarong et al.
Journal of virology, 93(2) (2018-11-02)
Influenza A virus (IAV), a major cause of human morbidity and mortality, continuously evolves in response to selective pressures. Stem-directed, broadly neutralizing antibodies (sBnAbs) targeting the influenza virus hemagglutinin (HA) are a promising therapeutic strategy, but neutralization escape mutants can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service