MAB8052
Anti-Adenovirus Antibody, clone 20/11
clone 20/11, Chemicon®, from mouse
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
clone
20/11, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
isotype
IgG1
shipped in
wet ice
General description
Adenoviruses are 65-80 nm, non-enveloped, regular icosahedron pathogens often associated with respiratory and gastrointestinal illness as well as conjunctivitis. Well over 40 types of adenoviruses have currently been recognized. In addition to their deleterious effects, adenoviruses have been used in vaccine production and in gene therapy. They also have the ability to trigger cell proliferation by interfering with host cellular anti-oncogenes. Since they are easy to prepare, multiple quickly and efficiently, and have the ability to infect and deliver their genome (including researcher-created constructs) to the nucleus of a variety of post-mitotic cells, adenoviruses are extremely valuable in research and medical applications ranging from virus/host cell interaction to the insertion of novel genes or regulatory mechanisms into eukaryotic cellular systems, to delivery of therapeutic anti-cancer genes.
Specificity
Recognizes All 41 serotypes of Adenovirus.
Immunogen
Adeno 3.
Application
Anti-Adenovirus Antibody, clone 20/11 detects level of Adenovirus & has been published & validated for use in ELISA, FC, IF, IH, IH(P).
EIA, IFA
Immunohistochemistry: 1:200. In trypsinized (with 0.1% trypsin in Tris buffer pH 7.6 containing 0.1% Calcium Cloride; for 5-10 minutes at room temp.) paraffin embedded tissue. Do not use antigen retrieval.
Flow cytometry.
Final working dilutions and protocols must be determined by end user.
Immunohistochemistry: 1:200. In trypsinized (with 0.1% trypsin in Tris buffer pH 7.6 containing 0.1% Calcium Cloride; for 5-10 minutes at room temp.) paraffin embedded tissue. Do not use antigen retrieval.
Flow cytometry.
Final working dilutions and protocols must be determined by end user.
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Infectious Diseases - Viral
Physical form
Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Protein A purified
Storage and Stability
Maintain at +2–8°C in convenient aliquots for up to 12 months. Do not store in a diluted format.
During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.
During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.
Analysis Note
Control
Adenovirus Control Slides, Catalogue Number 5009-5
Adenovirus Control Slides, Catalogue Number 5009-5
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Efficient internalization into low-passage glioma cell lines using adenoviruses other than type 5: an approach for improvement of gene delivery to brain tumours.
Skog, J; Edlund, K; Widegren, B; Salford, LG; Wadell, G; Mei, YF
The Journal of General Virology null
Adenovirus type 7 induces interleukin-8 in a lung slice model and requires activation of Erk.
Booth, JL; Coggeshall, KM; Gordon, BE; Metcalf, JP
Journal of virology null
Simultaneous occurrence of lymphoepithelial cysts, cytomegalovirus and mycobacterial infections in the intraparotid lymph nodes of a patient with AIDS.
P A Vargas, H Villalba, A P Passos, P H Saldiva, T Mauad, H H Caiaffa Filho, A Lucena, O P Almeida
Journal of Oral Pathology & Medicine null
David Marchant et al.
Journal of virology, 84(21), 11359-11373 (2010-08-13)
Respiratory viruses exert a heavy toll of morbidity and mortality worldwide. Despite this burden there are few specific treatments available for respiratory virus infections. Since many viruses utilize host cell enzymatic machinery such as protein kinases for replication, we determined
Adenovirus types 11p and 35 attach to and infect primary lymphocytes and monocytes, but hexon expression in T-cells requires prior activation.
Anna Segerman, Kristina Lindman, Ya-Fang Mei, Annika Allard, Goran Wadell
Virology null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service