Skip to Content
Merck
All Photos(1)

Key Documents

MAB5448

Sigma-Aldrich

Anti-Neurofilament H (200 kDa) Antibody

CHEMICON®, rat monoclonal, TA51

Synonym(s):

NF-H

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Neurofilament NF-H Antibody, clone TA51, culture supernatant, clone TA51, Chemicon®

biological source

rat

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

TA51, monoclonal

species reactivity

human, rat, hamster, monkey

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NEFH(4744)
rat ... Nefh(24587)
rhesus monkey ... Nefh(717705)

Specificity

Neurofilament H (NF-H), carboxy terminal (phosphate dependent (+). Clone is specific for phosphorylated NF-H {Carden et al (1987) 7(11):3489}.

Immunogen

Epitope: carboxy terminal phosphorylation dependent, within the multiphosphorylation repeat in NFH

Application

Anti-Neurofilament NF-H Antibody, clone TA51 is an antibody against Neurofilament NF-H for use in ELISA, IP, WB, IH.
Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism

Neuronal & Glial Markers
Western blot: Identifies ~ 200kDa band in reduced samples. Use Tris based buffers for best results as epitope is phosphate dependent; Different regions of the CNS were dissected out immediately and homogenized in BUST buffer (8 M urea, 0.5% SDS, 2% 13-mercaptoethanol, and 50 mM Tris, pH 7.4) at a ratio of i ml/100 mg wet tissue. The tissue suspensions were sonicated well and centrifuged in a TL-100 ultracentrifuge (Beckman Instruments, Fullerton, CA) at 100,000 g

for 1 h at 25°C, and the protein concentrations in the supernatants were determined. Proteins were separated on 7.5% SDS-PAGE gels, {Tu. P et al (1995) J Cell Biol 129:1629-1640}

Immunohistochemistry on frozen sections; not effective on paraffin tissues in original work. {Carden et al, 1987}.

Optimal working dilutions must be determined by end user.

Physical form

Unpurified tissue culture supernatant from a perfusion system, filtered through a 0.2μ membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natasha Sukhanov et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 41(7), 1393-1400 (2021-01-06)
Cell adhesion proteins of the Cadm (SynCAM/Necl) family regulate myelination and the organization of myelinated axons. In the peripheral nervous system (PNS), intercellular contact between Schwann cells and their underlying axons is believed to be mediated by binding of glial
Soundara Rajan Thangavelu et al.
Annals of neurosciences, 18(2), 48-55 (2011-04-01)
Amyotrophic Lateral Sclerosis, in which motor neurons degenerate, leading to paralysis, not only the affected motor neurons, but the surrounding non-neuronal cells also contribute significantly to the disease. However, the disease mechanism is not known. In this study we have
Jean Defourny et al.
Nature communications, 4, 1438-1438 (2013-02-07)
Hearing requires an optimal afferent innervation of sensory hair cells by spiral ganglion neurons in the cochlea. Here we report that complementary expression of ephrin-A5 in hair cells and EphA4 receptor among spiral ganglion neuron populations controls the targeting of
Bacterial neurotoxic metabolites in multiple sclerosis cerebrospinal fluid and plasma.
Ntranos, et al.
Brain, 145, 569-583 (2023)
Stimulation of adult oligodendrogenesis by myelin-specific T cells.
Hvilsted Nielsen, H; Toft-Hansen, H; Lambertsen, KL; Owens, T; Finsen, B
The American Journal of Pathology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service