Skip to Content
Merck
All Photos(3)

Key Documents

MAB5320

Sigma-Aldrich

Anti-Synuclein α Antibody, clone 7B12.2

culture supernatant, clone 7B12.2, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

7B12.2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SNCA(6622)

Specificity

Reacts with alpha Synuclein. By Western blot the antibody shows no reactivity with beta or gamma synuclein. This antibody was shown to stain Lewy bodies in frozen section of human brain with Parkinson′s disease and glial inclusions in frozen section of human brain with Multiple System Atrophy.

Immunogen

Recombinant human alpha synuclein.

Application

Anti-Synuclein Antibody, α, clone 7B12.2 detects level of Synuclein & has been published & validated for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western blotting on recombinant alpha synuclein: 1:2,000-1:4,000.

Lane 1: Recombinant human alpha synuclein

Lane 2: Recombinant human beta synuclein

Lane 3: Recombinant human gamma synuclein

Immunohistochemistry (frozen sections): 1:250-1:500

Optimal working dilutions must be determined by end user.

Target description

15-18 kDa

Linkage

Replaces: 04-1053

Physical form

UnPurified tissue culture supernatant from a perfusion system, filtered through a 0.2μ micron membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Sardone et al.
Matrix biology : journal of the International Society for Matrix Biology, 55, 90-105 (2016-10-19)
In response to injury, tendon fibroblasts are activated, migrate to the wound, and contribute to tissue repair by producing and organizing the extracellular matrix. Collagen VI is a microfibrillar collagen enriched in the pericellular matrix of tendon fibroblasts with a
Francesca Sardone et al.
Journal of cellular physiology, 229(7), 878-886 (2013-12-21)
Cell-extracellular matrix interaction plays a major role in maintaining the structural integrity of connective tissues and sensing changes in the biomechanical environment of cells. Collagen VI is a widely expressed non-fibrillar collagen, which regulates tissues homeostasis. The objective of the
Xichen Zheng et al.
The Journal of clinical investigation, 128(5), 2104-2115 (2018-04-18)
Immune checkpoint blockade (ICB) has demonstrated curative potential in several types of cancer, but only for a small number of patients. Thus, the identification of reliable and noninvasive biomarkers for predicting ICB responsiveness is an urgent unmet need. Here, we
Nanoscale imaging of individual amyloid aggregates extracted from brains of Alzheimer and Parkinson patients reveals presence of lipids in I?-synuclein but not in amyloid I?1-42 fibrils.
Zhaliazka, et al.
Protein Science, 32, e4598-e4598 (2023)
Xianjun Chen et al.
Frontiers in neuroscience, 15, 659853-659853 (2021-05-08)
Increasing evidence has demonstrated that in addition to dysfunction of neuronal circuitry, oligodendroglial dysfunction and/or disruption of white matter integrity are found in the brains of patients with schizophrenia. DNA methylation, a well-established risk factor for schizophrenia, has been demonstrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service