Skip to Content
Merck
All Photos(2)

Documents

MAB5256A5

Sigma-Aldrich

Anti-Neurofilament 200kDa Antibody, clone NE14, Alexa Fluor 555 Conjugate

clone NE14, from mouse, ALEXA FLUOR 555

Synonym(s):

Neurofilament heavy polypeptide, NF-H, 200 kDa neurofilament protein, Neurofilament triplet H protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 555

antibody form

purified antibody

antibody product type

primary antibodies

clone

NE14, monoclonal

species reactivity

mouse, rat, pig

species reactivity (predicted by homology)

human (100% sequence homology), porcine (immunogen homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NEFH(4744)
mouse ... Nefh(380684)
pig ... Nefh(100156492)
rat ... Nefh(24587)

General description

Neurofilaments are a type of intermediate filament that serve as major elements of the cytoskeleton supporting the axon cytoplasm. They are the most abundant fibrillar components of the axon, being on average 3-10 times more frequent than axonal microtubules. Neurofilaments (10nm in dia.) are built from three intertwined protofibrils which are themselves composed of two tetrameric protofilament complexs of monomeric proteins. The neurofilament triplet proteins (68/70, 160, and 200 kDa) occur in both the central and peripheral nervous system and are usually neuron specific. The 68/70 kDa NF-L protein can self-assemble into a filamentous structure, however the 160 kDa NF-M and 200 kDa NF-H proteins require the presence of the 68/70 kDa NF-L protein to co-assemble. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas and neuroblastomas stain positively for neurofilaments. Although typically restricted to neurons, neurofilaments have been detected in paragangliomas and adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilaments. For more neurofilament information and epitope maps, search "neurofilament" on our website.

Application

Detect Neurofilament 200kDa using this Anti-Neurofilament 200kDa Antibody, clone NE14, Alexa Fluor 555 Conjugate validated for use in ICC & IHC.

Quality

Evaluated by Immunocytochemistry in rat E18 primary cortex cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Neurofilament in rat E18 primary cortex cells.

Other Notes

Unconjugated Anti-Neurofilament has been shown to work with multiple species including Human and Porcine.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giulio Morozzi et al.
iScience, 24(12), 103434-103434 (2021-12-09)
Inflammatory responses are crucial for regeneration following peripheral nerve injury (PNI). PNI triggers inflammatory responses at the site of injury. The DNA-sensing receptor cyclic GMP-AMP synthase (cGAS) and its downstream effector stimulator of interferon genes (STING) sense foreign and self-DNA
Yongchen Cui et al.
Anesthesiology, 139(6), 782-800 (2023-09-05)
Continuous nerve block with ropivacaine is commonly performed after repair surgery for traumatic peripheral nerve injuries. After peripheral nerve injury, tetrodotoxin-resistant voltage-gated sodium channel Nav1.8 is upregulated and contributes to macrophage inflammation. This study investigated whether ropivacaine promotes peripheral nerve
X Zhao et al.
Oncogene, 35(27), 3565-3576 (2015-11-10)
Previous studies have indicated the important roles of MYCN in tumorigenesis and progression of neuroblastoma (NB), the most common extracranial solid tumor derived from neural crest in childhood. However, the regulatory mechanisms of MYCN expression in NB still remain largely
Sofía Ibarburu et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 18(1), 309-325 (2020-10-30)
Motor neuron degeneration and neuroinflammation are the most striking pathological features of amyotrophic lateral sclerosis (ALS). ALS currently has no cure and approved drugs have only a modest clinically therapeutic effect in patients. Drugs targeting different deleterious inflammatory pathways in
Marie-Pier Girouard et al.
eNeuro, 7(2) (2020-02-01)
In contrast to neurons in the CNS, damaged neurons from the peripheral nervous system (PNS) regenerate, but this process can be slow and imperfect. Successful regeneration is orchestrated by cytoskeletal reorganization at the tip of the proximal axon segment and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service