Skip to Content
Merck
All Photos(1)

Key Documents

MAB5230

Sigma-Aldrich

Anti-CASK Antibody, clone K56A

clone K56A, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

K56A, monoclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASK(8573)
rat ... Cask(29647)

Specificity

CASK. The immunogen protein represents a CASK-specific domain that lies between the calmodulin binding and PDZ domains. MAB5230 exhibits a specific reaction to the two alternatively spliced isoforms of CASK present in the mammalian CNS, the CASK-B or long (~900 amino acid) isoform present in rat brain membrane preparations, and the soluble CASK-A or short (~600 amino acid) isoform.CASK is a synapse-associated Ca2+/calmodulin-dependent protein kinase that also contains PDZ, SH3 and guanylate kinase domains. CASK is thought to play a critical role in synaptic vesicle exocytosis and synaptic junctions.

Immunogen

CASK-GST fusion protein (amino acids 318-415).

Application

Anti-CASK Antibody, clone K56A is an antibody against CASK for use in WB, IC, IH.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western blot: 1 μg/mL

Immunocytochemistry: 0.5-1.0 μg/mL

Immunohistochemistry on 4% paraformaldehyde fixed tissue: 0.5- 1.0 μg/mL

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Neupert et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(40), 13629-13647 (2015-10-09)
Synapses depend on trafficking of key membrane proteins by lateral diffusion from surface populations and by exocytosis from intracellular pools. The cell adhesion molecule neurexin (Nrxn) plays essential roles in synapses, but the dynamics and regulation of its trafficking are
Volker M Stucke et al.
Molecular biology of the cell, 18(5), 1744-1755 (2007-03-03)
Three groups of evolutionarily conserved proteins have been implicated in the establishment of epithelial cell polarity: the apically-localized proteins of the Par (Par3-Par6-aPKC-Cdc42) and Crumbs groups (Crb3-PALS1-PATJ) and the basolaterally localized proteins of the Dlg group (Dlg1-Scribble-Lgl). During epithelial morphogenesis
Hsiao-Tang Hu et al.
Scientific reports, 6, 33592-33592 (2016-09-16)
Functional synapse formation requires tight coordination between pre- and post-synaptic termini. Previous studies have shown that postsynaptic expression of heparan sulfate proteoglycan syndecan-2 (SDC2) induces dendritic spinogenesis. Those SDC2-induced dendritic spines are frequently associated with presynaptic termini. However, how postsynaptic
Aswini Gnanasekaran et al.
Journal of neurochemistry, 126(1), 102-112 (2013-04-23)
ATP-gated P2X3 receptors of sensory ganglion neurons are important transducers of painful stimuli and are modulated by extracellular algogenic substances, via changes in the receptor phosphorylation state. The present study investigated the role of calcium/calmodulin-dependent serine protein kinase (CASK) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service