Skip to Content
Merck
All Photos(4)

Key Documents

MAB4155

Sigma-Aldrich

Anti-BCRP1 Antibody, clone 5D3

clone 5D3, Chemicon®, from mouse

Synonym(s):

ABC transporter, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, ATP-binding cassette, sub-family G, member 2, Breast cancer resistance protein, CD338 antigen, Mitoxantrone resistance-associated protein, Placent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5D3, monoclonal

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

input

sample type neural stem cell(s)
sample type hematopoietic stem cell(s)

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BCRP1(644079)

Related Categories

General description

Breast cancer resistance protein (BCRP1), also known as placenta-specific ATP-binding cassette (ABC) protein (ABCP) or ABC G-subfamily member 2 (ABCG2), or mitoxantrone resistance protein (MXR), is a member of the ATP-binding cassette superfamily of drug transporters thought to be involved in multi-drug resistance in human neoplastic disease. ABCG2 is expressed in drug-resistant breast, colon and gastric cancer and fibrosarcoma cancer cell lines and in blast cells from patients with acute myeloid leukemia (AML). Normal tissues that express the ABCG2 protein include placental syncytiothrophoblasts, mammary ducts and lobules, epithelium in the large intestine and colon, venous and capillary epithelium and bile canalicular membrane of the liver. Several studies indicate that ABCG2 is also expressed in stem cells of different tissues and may be useful as a stem cell marker. ABCG2 is thought to be responsible for the ability of a population of very primitive hematopoietic and non-hematopoietic cells, the so-called SP (side population) subset, to efflux the Hoechst 33342 fluorescent dye. Semiquantitative RT-PCR results indicate that ABCG2 mRNA is expressed at higher levels in SP cells as compared to non-SP cells in human, rhesus monkey and mouse hematopoietic tissues, with the exception of erythroblasts and NK cells. In human bone marrow, ABCG2 mRNA expression is higher in phenotypically defined populations of primitive CD34+CD38- hematopoietic cells as compared to more differentiated CD34+CD38+ progenitor cells. Bcrp1 mRNA is also expressed in SP cells in murine skeletal muscle, in neural stem cell and progenitor cells and in nestin-positive pancreatic islet derived progenitor cells. Over-expression of the human ABCG2 gene by retroviral-mediated gene transfer confers the SP phenotype to mouse bone marrow cells. Bcrp1 mRNA is expressed in SP as well as non-SP fractions of mouse embryonic stem cells. This suggests that Bcrp1 protein levels may not always be reflected by mRNA levels, or that other characteristics of stem cells, for example, cell cycle status or expression of other transporters, are also involved in the SP phenotype.

Specificity

Specifically recognizes an external epitope of the human form of ABCG2. Reportedly reacts with mouse stem cell lines (Ginis, et al., 2004), however mouse reactivity is unsupported at this time.

Immunogen

3T3 cells expressing human ABCG2

Application

Detect BCRP1 using this Anti-BCRP1 Antibody, clone 5D3 validated for use in FC, IC, IF, FUNC & WB.
Immunocytochemistry:
Cell cultures fixed with 4% PFA 5-15 minutes at room temperature. Blocking with 10% NGS, 0.1% BSA in PBS with 0.3% Triton X-100 (Ginis et al., 2004).

Flow Cytometry:
1:100-1:1000 dilution from a previous lot was used.

Inhibition Assay:
A previous lot was used to inhibit ABCG2 (Watanabe, K. et al., (2004. FEBS Letters 565:6-10). See also Sarkadi, B., et al., 2004 FEBS 567:116-120,. Sarkadi reports that the detection and inhibition by 5D3 strongly depends on the actual conformation of ABCG2.
Blocking with 10% NGS, 0.1% BSA in PBS with 0.3% triton X-100 (Ginis, 2004).

Optimal working dilutions must be determined by end user.
Research Category
Stem Cell Research
Research Sub Category
Neural Stem Cells

Hematopoietic Stem Cells

Quality

Routinely evaluated by Western Blot on L6 lysates.

Western Blot Analysis:
1:500 dilution of this lot detected BCRP1 on 10 μg of L6 lysates.

Target description

72 kDa

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG2bκ liquid in buffer containing PBS, pH 7.4, with 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt. Do not freeze.

Analysis Note

Control
MCF-7 cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenji Zhang et al.
The Journal of biological chemistry, 294(52), 20222-20232 (2019-11-14)
ATP-binding cassette (ABC) subfamily G member 2 (ABCG2) belongs to the ABC transporter superfamily and has been implicated in multidrug resistance of cancers. Although the structure and function of ABCG2 have been extensively studied, little is known about its biogenesis
A Khammanivong et al.
Veterinary and comparative oncology, 14(3), e113-e125 (2014-08-13)
Canine hemangiosarcoma is a rapidly progressive disease that is poorly responsive to conventional chemotherapy. Despite numerous attempts to advance treatment options and improve outcomes, drug resistance remains a hurdle to successful therapy. To address this problem, we used recently characterized
Comparative proteomics reveals human pluripotent stem cell-derived limbal epithelial stem cells are similar to native ocular surface epithelial cells.
Mikhailova, A; Jylha, A; Rieck, J; Nattinen, J; Ilmarinen, T; Vereb, Z; Aapola et al.
Scientific Reports null
Christophe Antczak et al.
Assay and drug development technologies, 12(1), 28-42 (2013-09-03)
ABCG2 is a member of the ATP-binding cassette (ABC) family of transporters, the overexpression of which has been implicated in resistance to various chemotherapeutic agents. Though a number of cell-based assays to screen for inhibitors have been reported, they do
Megan H Cox et al.
The Biochemical journal, 475(9), 1553-1567 (2018-04-18)
Multidrug binding and transport by the ATP-binding cassette transporter ABCG2 is a factor in the clinical resistance to chemotherapy in leukaemia, and a contributory factor to the pharmacokinetic profiles of many other prescribed drugs. Despite its importance, the structural basis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service