Skip to Content
Merck
All Photos(1)

Key Documents

MAB352

Sigma-Aldrich

Anti-Serotonin Antibody, clone YC5/45

culture supernatant, clone YC5/45, Chemicon®

Synonym(s):

5-HT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

YC5/45, monoclonal

species reactivity

mouse, human, rat, lizard, rabbit, pig

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgG

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... HTR1A(3350)

Specificity

Recognizes the formaldehyde conjugate of serotonin. Without paraformaldehyde treatment, it cross-reacts in liquid phase with dopamine and tryptamine in hemagglutination tests. Shows no reactivity with catecholamine-containing neurons in formaldehyde fixed preparations. MAB352 reacts with serotonin in neurons and recognizes serotonergic sites in fixed tissue sections. By cell agglutination, no cross reactivity with 5-hydroxyindolacetic acid, GABA, noradrenaline, 5-hydroxytryptophan, carnosine or melatonin.

Immunogen

Serotonin conjugated to BSA.

Application

Anti-Serotonin Antibody, clone YC5/45 is an antibody against Serotonin for use in IH.
Immunohistochemistry at 1:100-1:200 for formaldehyde fixed tissue.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Lyophilized Supernatant, preservative free.
Unpurified

Storage and Stability

Lyophilized antibody can be kept at 2-8°C for up to 3 months and should be kept at -20°C for long term storage. Reconstitute with 100μL sterile DI water. Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze-thaw cycles.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Drobyshevsky et al.
Journal of neurochemistry, 132(4), 394-402 (2014-11-26)
We hypothesized that a deficiency in the descending serotonergic input to spinal cord may underlie postnatal muscle hypertonia after global antenatal hypoxic-ischemic injury in a rabbit model of cerebral palsy. Neurotransmitter content was determined by HPLC in the spinal cord
Tanimul Alam et al.
Nature communications, 7, 10388-10388 (2016-01-23)
The molecular mechanisms underlying the ability of axons to regenerate after injury remain poorly understood. Here we show that in Caenorhabditis elegans, axotomy induces ectopic expression of serotonin (5-HT) in axotomized non-serotonergic neurons via HIF-1, a hypoxia-inducible transcription factor, and
Carlos Pantoja et al.
Neuron, 91(3), 587-601 (2016-07-12)
Inter-individual behavioral variation is thought to increase fitness and aid adaptation to environmental change, but the underlying mechanisms are poorly understood. We find that variation between individuals in neuromodulatory input contributes to individuality in short-term habituation of the zebrafish (Danio
Basudha Basu et al.
Reproduction (Cambridge, England), 135(5), 657-669 (2008-02-29)
Serotonin is reported to be present in early embryos of many species and plays an important role in early patterning. Since it is a fluorophore, it can be directly visualized using fluorescence microscopy. Here, we use three-photon microscopy to image
Yu-Chia Chen et al.
The Journal of biological chemistry, 291(42), 21880-21892 (2016-08-20)
Hypothalamic neurons expressing histamine and orexin/hypocretin (hcrt) are necessary for normal regulation of wakefulness. In Parkinson's disease, the loss of dopaminergic neurons is associated with elevated histamine levels and disrupted sleep/wake cycles, but the mechanism is not understood. To characterize

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service