Skip to Content
Merck
All Photos(4)

Key Documents

MAB3303

Sigma-Aldrich

Anti-Collagen Type VI Antibody, clone VI-26

clone VI-26, Chemicon®, from mouse

Synonym(s):

Anti-BTHLM1, Anti-OPLL, Anti-UCHMD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

VI-26, monoclonal

species reactivity

human, rabbit

should not react with

rat

manufacturer/tradename

Chemicon®

availability

not available in Japan

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... COL6A1(1291)

Specificity

Specifically reacts with the hCL(VI). This is a purified mouse monoclonal antibody to human type VI collagen. VI-26 does not react with denatured or reduced type VI collagen, although this antibody does recognize the the triple helix consisting of alpha1(VI), alpha 2(VI) alpha3(gamma). The epitope has not been mapped and the monoclonal is know not to react with collagens I, II, III, IV or V in native assays.

Application

Anti-Collagen Type VI Antibody, clone VI-26 detects level of Collagen Type VI & has been published & validated for use in ELISA, IH(P).
Immunohistochemistry on acetone fixed paraffin-embedded tissues. Formalin fixation is not recommended. Antigen recovery is treatment with 0.04% trypsin in 0.01M CaCl(2) 0.05M Tris-HCL pH 7.6, 37°C for 10 minutes before the non-specific peroxidase block.

EIA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Format: Purified
Liquid in 0.1 M sodium phosphate buffer, pH 7.0, containing 2% protease-free bovine Serum albumin.
Protein A purified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Testis, connective tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiziana Triulzi et al.
PloS one, 8(2), e56761-e56761 (2013-02-27)
We recently showed that differential expression of extracellular matrix (ECM) genes delineates four subgroups of breast carcinomas (ECM1, -2, -3- and -4) with different clinical outcome. To further investigate the characteristics of ECM signature and its impact on tumor progression
D O Visscher et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 107(5), 1711-1721 (2018-11-02)
The aim of this study was to design and manufacture an easily assembled cartilage implant model for auricular reconstruction. First, the printing accuracy and mechanical properties of 3D-printed poly-ε-caprolactone (PCL) scaffolds with varying porosities were determined to assess overall material
Laura K Zamurs et al.
The Journal of biological chemistry, 290(7), 4272-4281 (2014-12-24)
Bethlem myopathy and Ullrich congenital muscular dystrophy (UCMD) sit at opposite ends of a clinical spectrum caused by mutations in the extracellular matrix protein collagen VI. Bethlem myopathy is relatively mild, and patients remain ambulant in adulthood while many UCMD
Laura Briñas et al.
Annals of neurology, 68(4), 511-520 (2010-10-27)
Mutations in the genes encoding the extracellular matrix protein collagen VI (ColVI) cause a spectrum of disorders with variable inheritance including Ullrich congenital muscular dystrophy, Bethlem myopathy, and intermediate phenotypes. We extensively characterized, at the clinical, cellular, and molecular levels
Russell J Butterfield et al.
Human mutation, 34(11), 1558-1567 (2013-09-17)
Glycine substitutions in the conserved Gly-X-Y motif in the triple helical (TH) domain of collagen VI are the most commonly identified mutations in the collagen VI myopathies including Ullrich congenital muscular dystrophy, Bethlem myopathy, and intermediate (INT) phenotypes. We describe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service