Skip to Content
Merck
All Photos(1)

Key Documents

MAB312

Sigma-Aldrich

Anti-A2B5 Antibody, clone A2B5-105

clone A2B5-105, Chemicon®, from mouse

Synonym(s):

Neuron Cell Surface Antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

A2B5-105, monoclonal

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgM

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Reacts with a surface antigen on neurons in the retina, brain, spinal cord and dorsal root ganglia, and with all tested human neuroblastoma cell lines. Does not seem to bind with any leukemic cell lines or with bone marrow cells from patients with leukemia, nor with cells from normal bone marrow. Cytotoxic to neurons in the presence of guinea-pig complement. Protein A binding is positive. Binds with GQ ganglioside on the plasma membrane of neurons of animals of all species tested to date (Eisenbarth, et al., 1979).

Immunogen

Embryonic chicken retinal cells

Application

Detect A2B5 using this Anti-A2B5 Antibody, clone A2B5-105 validated for use in FC, IC, IF, IH.
Indirect immunofluorescence (1:100 - 1:500) in the detection of neurons in tissue culture.

Using double immunofluorescence the GQ ganglioside has a similar distribution on neurons to the D2 protein, to the tetanus toxin receptor and to neurofilaments (Walsh, 1980).

May also be used in the depletion of neurons for a mixed population or their purification by affinity chromatography.

May also be useful for detecting the metastatic spread of neuroblastoma cells into bone marrow.

Flow cytometry: live cells {Maric, D. et al. (2000) Cerebral Cortex 10:729-747}.

Immunohistochemistry: Frozen, fixed tissues (Levison & McCarthy, 1989)

Complement-mediated cytotoxicity (Eisenbarth et al., 1979)

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Physical form

Format: Purified
Liquid in 0.02M PB, 0.25M NaCl, pH 7.6 containing 0.1% sodium azide.

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Type II astrocytes, human neural progenitors

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noah M Walton et al.
Development (Cambridge, England), 133(18), 3671-3681 (2006-08-18)
The isolation and expansion of human neural cell types has become increasingly relevant in restorative neurobiology. Although embryonic and fetal tissue are frequently envisaged as providing sufficiently primordial cells for such applications, the developmental plasticity of endogenous adult neural cells
Natalie D Bull et al.
Investigative ophthalmology & visual science, 50(9), 4244-4253 (2009-04-10)
Glaucoma is a common neurodegenerative disease for which current therapies are often insufficient; thus, new neuroprotective strategies are an important goal. Stem cells are attracting increasing attention as mediators of neuroprotection, often conferred via the trophic support of injured neurons.
Zheng Chen et al.
Scientific reports, 9(1), 9437-9437 (2019-07-03)
Accumulation of iron has been associated with the pathobiology of various disorders of the central nervous system. Our previous work has shown that hephaestin (Heph) and ceruloplasmin (Cp) double knockout (KO) mice induced iron accumulation in multiple brain regions and
Brittney A Beyer et al.
Nature chemical biology, 14(1), 22-28 (2017-11-14)
Endogenous metabolites play essential roles in the regulation of cellular identity and activity. Here we have investigated the process of oligodendrocyte precursor cell (OPC) differentiation, a process that becomes limiting during progressive stages of demyelinating diseases, including multiple sclerosis, using
Hala Gabr et al.
Cell transplantation, 24(9), 1813-1827 (2014-09-10)
Spinal cord injury (SCI) results in demyelination of surviving axons, loss of oligodendrocytes, and impairment of motor and sensory functions. We have developed a clinical strategy of cell therapy for SCI through the use of autologous bone marrow cells for

Articles

Human iPSC neural differentiation media and protocols used to generate neural stem cells, neurons and glial cell types.

Human iPSC neural differentiation media and protocols used to generate neural stem cells, neurons and glial cell types.

Human iPSC neural differentiation media and protocols used to generate neural stem cells, neurons and glial cell types.

Human iPSC neural differentiation media and protocols used to generate neural stem cells, neurons and glial cell types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service