MAB2511
Anti-VCAM-1 Antibody, clone P8B1
clone P8B1, Chemicon®, from mouse
Synonym(s):
CD106
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
P8B1, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
isotype
IgG2b
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... L1CAM(3897) , VCAM1(7412)
Specificity
MAB2511 recognizes the human VCAM-1 molecule. The antibody inhibits VLA-4 (integrin alpha4beta1) binding and recognizes a different epitope than that which is recognized by MAB2144 (P3C4 clone).
Application
Anti-VCAM-1 Antibody, clone P8B1 detects level of VCAM-1 & has been published & validated.
Function blocking
Optimal working dilutions must be determined by end user.
Optimal working dilutions must be determined by end user.
Physical form
Format: Purified
Purified immunoglobulin. Liquid in 0.2 M phosphate buffer to 0.25 M NaCl, pH 7.6 with 0.1% sodium azide. Caution: Sodium azide is toxic. See online MSDS.
Storage and Stability
Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.
During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.
During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thomas D Manes et al.
Arteriosclerosis, thrombosis, and vascular biology, 36(9), 1910-1918 (2016-07-23)
Circulating human T effector memory cell (TEM) recognition of nonself MHC (major histocompatibility complex) molecules on allograft endothelial cells can initiate graft rejection despite elimination of professional antigen-presenting cells necessary for naive T-cell activation. Our previous studies of CD4 TEM
Barbara Garmy-Susini et al.
The Journal of clinical investigation, 115(6), 1542-1551 (2005-05-20)
Neovascularization depends on vascular cell proliferation and on the stabilization of vessels by association of vascular smooth muscle-like pericytes with ECs. Here we show that integrin alpha4beta1 (VLA-4) and VCAM-1 promote close intercellular adhesion between ECs and pericytes and that
Max L B Grönloh et al.
EMBO reports, 24(1), e55483-e55483 (2022-11-17)
Upon inflammation, leukocytes leave the circulation by crossing the endothelial monolayer at specific transmigration "hotspot" regions. Although these regions support leukocyte transmigration, their functionality is not clear. We found that endothelial hotspots function to limit vascular leakage during transmigration events.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service