Skip to Content
Merck
All Photos(1)

Key Documents

MAB1976H

Sigma-Aldrich

Anti-Integrin αVβ3 Antibody, clone LM609, Phycoerythrin conjugated

clone LM609, Chemicon®, from mouse

Synonym(s):

Vitronectin Receptor, CD51/CD61

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LM609, monoclonal

species reactivity

monkey, pig, rabbit, bovine, avian, chicken, canine, human

should not react with

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

Related Categories

General description

EMISSION MAXIMUM:

580 nm
The integrin family of cell adhesion receptors consists of at least 16 membrane-associated heterodimers, composed of an alpha and beta subunit that associate in a non-covalent manner. The structure and functional diversity of the integrin family are based upon the pairing abilities of the individual alpha and beta subunits. Key to these molecular interactions between the integrin receptors and their respective ligands is the recognition of the Arg-Gly-Asp (RGD) sequence, known to be present in the extracellular matrix components fibronectin, vitronectin, collagen, fibrinogen, and von Willebrand factor (Cheresh, 1991). The involvement of integrins in vascular proliferation, adhesion, and wound repair has been well documented. The adhesion receptor, integrin alphaVbeta3, appears to be selectively expressed on growing blood vessels and has been identified as a marker of angiogenic vascular tissue (Brooks, 1994). Due to its involvement in angiogenesis, integrin alphaVbeta3 is one of the most intensely studied of the integrin receptors.

Specificity

Monoclonal antibody LM609 is reactive with the vitronectin receptor alphaVbeta3 complex, an RGD-directed adhesion receptor. LM609 has been demonstrated to block adhesion of a human melanoma cell line (M21) to vitronectin, fibrinogen and von Willebrand factor, as well as to a synthetic RGD containing peptide (Cheresh, 1987). In chick chorioallentoic membranes, LM609 was shown to block angiogenesis induced by bFGF and TNFalpha but had no effect on pre-existing vessels (Brooks, 1994). While LM609 does block cell attachment to RGD containing ligands, it does not interact directly with the RGD binding site. Instead, LM609 appears to be an allosteric inhibitor of integrin alphaVbeta3, which binds to a conformational epitope resulting from the post-translational association of the alphaV and beta3 subunits.

ABSORPTION MAXIMA: 90 nm, 545 nm and 565 nm

Immunogen

Purified adhesion receptor from the human melanoma cell line M21 (Cheresh, 1987).

Application

Anti-Integrin αVβ3 Antibody, clone LM609, Phycoerythrin conjugated detects level of Integrin αVβ3 & has been published & validated for use in FC, IH.
Flow Cytometry: 1-3 μg LM609 per 0.5 x 10e6 cells in 100μL (Smith, 1996).

Immunofluorescence: 5-10 μg/mL for staining of bFGF-treated 6 mm cryosections of chick chorioallantoic membrane fixed with acetone (Brooks, 1994). LM609 is also effective on 4% paraformaldehyde fixed, frozen tissues and cells. However it is not effective for immunohistochemical staining of paraffin-embedded tissue sections.

Optimal working dilutions and protocols must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Liquid in 0.01M sodium phosphate (pH7.6), 0.25 M NaCl, 0.05% sodium azide, 0.01% Thimerosal and 15 mg/mL BSA added as a stabilizer.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months. Protect from Light. Do not store as a dilute solution.

Analysis Note

Control
POSITIVE CONTROL: Human myeloma cell lines M21 and MoalphaV (Chen, 1995).

NEGATIVE CONTROL: Mo cells (this cell line was derived from M21 but does not express integrin alphaV; Chen, 1995).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Gandhi et al.
Molecular pharmaceutics, 6(2), 641-647 (2009-09-01)
Therapeutic proteins have emerged as a significant class of pharmaceutical agents over the past several decades. The potency, rapid elimination, and systemic side effects have prompted the need of spatiotemporally controlled release for proteins maybe more than any other active
Ellen Krautkrämer et al.
Virology journal, 9, 299-299 (2012-12-01)
Epithelio- and endotheliotropic viruses often exert polarized entry and release that may be responsible for viral spread and dissemination. Hantaviruses, mostly rodent-borne members of the Bunyaviridae family infect epithelial and endothelial cells of different organs leading to organ dysfunction or
Keren Cohen et al.
Oncotarget, 5(15), 6312-6322 (2014-07-30)
Thyroid hormone (3,5,3'-triiodothyronine, T3; L-thyroxine, T4) enhances cancer cell proliferation, invasion and angiogenesis via a discrete receptor located near the RGD recognition site on αvβ3 integrin. Tetraiodothyroacetic acid (tetrac) and its nanoparticulate formulation interfere with binding of T3/T4 to the
M Żmigrodzka et al.
Polish journal of veterinary sciences, 19(2), 407-414 (2016-08-04)
Platelets play a crucial role in hemostasis. Their activation has not yet been evaluated in healthy dogs with a normal and low platelet count. The aim of this study was to determine the influence of activators on platelet activation in
R S Watson et al.
Gene therapy, 20(6), 670-677 (2012-11-16)
With the long-term goal of developing a gene-based treatment for osteoarthritis (OA), we performed studies to evaluate the equine joint as a model for adeno-associated virus (AAV)-mediated gene transfer to large, weight-bearing human joints. A self-complementary AAV2 vector containing the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service