Skip to Content
Merck
All Photos(1)

Key Documents

MAB1613

Sigma-Aldrich

Anti-Kinesin Antibody, heavy chain, non-neuronal KHC, clone H1

clone H1, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

H1, monoclonal

species reactivity

rat, human, bovine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KIF5B(3799)

Specificity

Reacts with the major form of heavy chain of non-neuronal KHC near the N-terminus.

Immunogen

Epitope: heavy chain, non-neuronal KHC

Application

Anti-Kinesin Antibody, heavy chain, non-neuronal KHC, clone H1 detects level of Kinesin & has been published & validated for use in ELISA, IP, RIA, WB, IC, IH.
Immunohistochemistry on paraformaldehyde fixed tissue. Also works for EM.

Immunocytochemistry: paraformaldehyde fixed cells; 5-10′ RT, freshly made 4% PFA; permeabilize with 0.1-.2% triton X-100 in 3% normal serum blocking buffer

Immunoblotting: recognizes 124kDa KHC from non-neuronal samples.

Immunoprecipitation

Inhibition of kinesin mediated transport

EIA/RIA

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Liquid in 0.02 M Phosphate buffer, 0.25 M NaCl.pH 7.6 with 0.1% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sten Strunze et al.
Cell host & microbe, 10(3), 210-223 (2011-09-20)
Many viruses deliver their genomes into the host cell nucleus for replication. However, the size restrictions of the nuclear pore complex (NPC), which regulates the passage of proteins, nucleic acids, and solutes through the nuclear envelope, require virus capsid uncoating
Wen-Jun Wang et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(1), 62-69 (2013-10-12)
Previous studies identified a family of organic anion transport proteins (OATPs), many of which have C-terminal PDZ binding consensus sequences. In particular, the C-terminal four amino acids of Oatp1a1, a transporter on rat and mouse hepatocytes, comprise a consensus binding
Souvik Sarkar et al.
Traffic (Copenhagen, Denmark), 7(8), 1078-1091 (2006-06-01)
Intracellular trafficking regulates the abundance and therefore activity of transporters present at the plasma membrane. The transporter, Na+-taurocholate co-transporting polypeptide (ntcp), is increased at the plasma membrane upon treatment of cells with cAMP, for which microtubules (MTs) are required and
Jie Zhou et al.
PLoS pathogens, 14(5), e1007055-e1007055 (2018-05-22)
Many viruses, including adenovirus, exhibit bidirectional transport along microtubules following cell entry. Cytoplasmic dynein is responsible for microtubule minus end transport of adenovirus capsids after endosomal escape. However, the identity and roles of the opposing plus end-directed motor(s) remain unknown.
Hongtao Chen et al.
Biophysical journal, 94(4), 1508-1520 (2007-11-06)
The role of cholesterol in the regulation of endosome motility was investigated by monitoring the intracellular trafficking of endocytosed folate receptors (FRs) labeled with fluorescent folate conjugates. Real-time fluorescence imaging of HeLa cells transfected with green fluorescent protein-tubulin revealed that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service