KK-10L-BK
Intermediates for Further Manufacturing Use (FFMU) - RHD (Rh) Specificities
Anti-D from the clone MS-26, Human IgG, Intermediate for further manufacturing use
Synonym(s):
Anti-D (clone MS-26), FFMU
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116102
NACRES:
NA.41
Recommended Products
biological source
human
Quality Level
antibody form
unpurified
clone
MS-26, monoclonal
form
liquid
packaging
pack of 10 L
concentration
1-10 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
6.7-7.7
isotype
IgG
shipped in
ambient
storage temp.
2-8°C
General description
Standardized, concentrated cell culture supernatant which has been diafiltered with phosphate buffered saline.
For the formulation of Anti-D blood typing reagents or devices.
This antibody reacts with DVI red cells. Since it is IgG, a sensitisation technique is required to induce agglutination, e.g.: anti-human globulin, enzyme addition or by several automated methodologies. It is an ideal blending partner with any of the Millipore IgM Anti-D FFMUs to create an IgM/IgG blended reagent.
Contains 0.1% (w/v) Sodium Azide; Shelf Life: 24 months from date of filtration.
Licensed by the US FDA under a shared manufacturing agreement.
For the formulation of Anti-D blood typing reagents or devices.
This antibody reacts with DVI red cells. Since it is IgG, a sensitisation technique is required to induce agglutination, e.g.: anti-human globulin, enzyme addition or by several automated methodologies. It is an ideal blending partner with any of the Millipore IgM Anti-D FFMUs to create an IgM/IgG blended reagent.
Contains 0.1% (w/v) Sodium Azide; Shelf Life: 24 months from date of filtration.
Licensed by the US FDA under a shared manufacturing agreement.
Application
Antibody intermediate for further manufacturing use.
Production of an in vitro blood typing product or device.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Partially processed reagent for further manufacture of non-injectable products only. The absence of all viral antigens has not been determined. No US Standard of Potency. Do not use if turbid. Contains 0.1%<nbsp/>(w/v) sodium azide.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service