Skip to Content
Merck
All Photos(2)

Key Documents

JGWP09025

Millipore

PTFE Membrane Filter, 0.2 μm Pore Size

Omnipore, filter diam. 90 mm, hydrophilic

Synonym(s):

Omnipore Membrane Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

material

PTFE membrane
plain filter
white filter

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Millipore
Omnipore

parameter

130 °C max. temp.

diam.

90 mm

filter diam.

90 mm

thickness

65 μm

pore size

0.2 μm pore size
80 % porosity

shipped in

ambient

General description

Omnipore membrane is a hydrophilic polytetrafluoroethylene (PTFE) membrane that is applicable for the filtration of aqueous solutions, clarifying acids, and alkaline solutions.

Application

  • Filtration of aqueous solutions
  • Clarifying acidic and alkaline solutions
  • HPLC, UPLC, LC-MS/MS mobile phase filtration
  • Microplastics analysis

Features and Benefits

  • Biologically and chemically inert.
  • High porosity yields high flow rates.
  • Compatible with a variety of solvents, acids, and alkaline solution.

Legal Information

Omnipore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Barbara Bernardim et al.
Nature protocols, 14(1), 86-99 (2018-11-25)
There is considerable interest in the development of chemical methods for the precise, site-selective modification of antibodies for therapeutic applications. In this protocol, we describe a strategy for the irreversible and selective modification of cysteine residues on antibodies, using functionalized
Ammonia synthesis at intermediate temperatures in solid-state electrochemical cells using cesium hydrogen phosphate based electrolytes and noble metal catalysts
Kishira S, et al.
International Journal of Hydrogen Energy, 42(43), 26843-26854 null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service