IPAKQUA00
IPAK Quanta® Polishing Cartridge for Purified Water
For Milli-Q® IQ/EQ 7000 and EQ 7008 7016 ultrapure water systems fed with purified water. This product replaces IPAKQUAA1.
About This Item
Recommended Products
Quality Level
packaging
pkg of 1 unit
manufacturer/tradename
Milli-Q®
humidity measuring range
10-80% (In original sealed packaging to protect purification elements)
H
39.5 cm (15.5 in.)
input
feed water nature purified water
compatibility
for use with Milli-Q® EQ 7000
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® IQ 7000
shipped in
ambient
storage temp.
2-30°C
Related Categories
General description
Combined with the IPAK Meta® polishing cartridge for purified water (IPAKMET00), it enables the production of ultrapure (Type 1) water for sensitive laboratory applications.
Application
- For Milli-Q® IQ/EQ 7000 and EQ 7008/16 ultrapure water purification systems fed with purified water
- Delivers ultrapure (Type 1) water for sensitive applications
Features and Benefits
- e-Sure tag enables RFID connection with the system, for seamless consumable status monitoring, data management and full traceability
- Supplied with a Drain Cap, to remove excess water and reduce the weight of the exhausted cartridge at disposal
- 78% of the parts suppliers are ISO 14001 environmental management system-certified
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.
Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.
Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.
Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service