Skip to Content
Merck

IP05

Protein G Plus/Protein A Agarose Suspension

Protein G PLUS/Protein A-Agarose mixture specifically formulated for immunoprecipitation.

Synonym(s):

Affinity resin, Protein G/Agarose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.56
UNSPSC Code:
41116133
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

slurry (Liquid)

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

technique(s)

protein purification: suitable

suitability

suitable for microbiology

shipped in

wet ice

storage temp.

2-8°C

General description

Designed for immunoprecipitation applications. This product is blocked with BSA to reduce non-specific binding and cannot be used for purification.
Protein G PLUS/Protein A-Agarose mixture specifically formulated for immunoprecipitation.

Application

Immunoprecipitation (see comments)

Physical form

33% slurry in PBS.

Other Notes

Agarose solution is supplied ready to use. Protein G Plus/Protein A Agarose immunoprecipitation reagent is blocked with BSA and should not be used for immunoglobulin purification or covalent cross-linking. For immunoprecipitation reactions 15 µl of solution per µg primary antibody is recommended. Preclearing will minimize extra bands resulting from nonspecific precipitation. To preclear, add to the sample 20 µl of agarose conjugate and 1 µg of normal IgG from the same species as the immunoprecipitating antibody. When immunoblotting is used for detection, some secondary antibodies can react nonspecifically with BSA or other proteins present at high concentrations in the sample. This can be eliminated by reducing the concentration of secondary antibody.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Abdulaziz Asiri et al.
Pathology international, 69(7), 381-391 (2019-07-11)
Cten is an oncogene promoting EMT in many signaling pathways, namely through Snail. We investigated whether Cten function could be mediated through Src. Cten levels were modulated by forced expression in HCT116 and gene knockdown in SW620 CRC (colorectal cancer)
Lianjun Zhang et al.
Nature communications, 12(1), 6154-6154 (2021-10-24)
Acute myeloid leukemia (AML) harboring inv(16)(p13q22) expresses high levels of miR-126. Here we show that the CBFB-MYH11 (CM) fusion gene upregulates miR-126 expression through aberrant miR-126 transcription and perturbed miR-126 biogenesis via the HDAC8/RAN-XPO5-RCC1 axis. Aberrant miR-126 upregulation promotes survival
Kylene Kehn et al.
Retrovirology, 1, 6-6 (2004-06-01)
The human T-cell leukemia virus type 1 (HTLV-1) Tax protein indirectly influences transcriptional activation, signal transduction, cell cycle control, and apoptosis. The function of Tax primarily relies on protein-protein interactions. We have previously shown that Tax upregulates the cell cycle



Global Trade Item Number

SKUGTIN
IP05-1.5ML04055977220223