Skip to Content
Merck
All Photos(1)

Key Documents

HTS191RTA

Sigma-Aldrich

Ready-to-Assay OXGR1 α-Ketoglutarate Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

Product Name

Ready-to-Assay OXGR1 α-Ketoglutarate Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

OXGR1 (also known as GPR99 or GPR80) was originally identified as an orphan with a sequence most closely related to P2Y receptors. Studies to deorphanize OXGR1 revealed that α-ketoglutarate activates OXGR1 through Gq to increase intracellular calcium (He et al., 2004; Qi et al., 2004; Abbracchio et al., 2005). The EC50 of OXGR1 for α-ketoglutarate is in the μM range, similar to the concentration of α-ketoglutarate in the circulation, indicating that the interaction is physiologically significant. Millipore′s cloned human OXGR1-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant OXGR1-expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists, and modulators at OXGR1.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human OXGR1, GPR99, GPR80 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: OXGR, GPR99, GPR80
  • Target Sub-Family: alpha-Ketoglutarate

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service