Skip to Content
Merck
All Photos(1)

Key Documents

HTS011RTA

Sigma-Aldrich

Ready-to-Assay CCR6 Chemokine Family Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.84

Product Name

Ready-to-Assay CCR6 Chemokine Family Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

CCR6 is a GPCR that binds to a single chemokine, CCL20 (also known as MIP-3α, LARC or Exods-1), as well as several members of the beta-defensin family of antimicrobial peptides (Schutyser, 2003). Immature dendritic cells, B-cells, and effector/memory T-cells express CCR6, and the receptor promotes inflammation during the development of rheumatoid arthritis, inflammatory bowel disease, and graft-versus-host disease (Varona et al., 2003; Kleinewietfeld et al., 2005; Varona et al., 2005). Millipore′s cloned human CCR6-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant CCR6 expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at CCR6.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Human CCR6 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: CCR6
  • Target Sub-Family: Chemokine

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service