Skip to Content
Merck
All Photos(2)

Key Documents

HNCSMAG-35K

Millipore

MILLIPLEX® Human Circadian/Stress Magnetic Bead Panel - Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

species reactivity

human

Quality Level

manufacturer/tradename

Milliplex®

assay range

accuracy: 129%
(Spike Recovery for Melatonin)

sensitivity: 4.3 pg/mL
(Melatonin; MinDC+2SD)

sensitivity: 686 pg/mL
(Cortisol; MinDC+2SD)

standard curve range: 3.4-2,500 pg/mL
(Melatonin)

standard curve range: 686-500,000 pg/mL
(Cortisol)

inter-assay cv: <25%
intra-assay cv: <15%

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

The MILLIPLEX® Human Circadian/Stress Bead Panel enables you to focus on the relevance and therapeutic potential of Melatonin and Cortisol in your research model. Coupled with the Luminex® xMAP® platform in a bead format, you receive the advantage of ideal speed and sensitivity, allowing quantitative multiplex detection of multiple analytes simultaneously, which can dramatically improve productivity.

The MILLIPLEX® Human Circadian/Stress Bead Panel is to be used for the simultaneous quantification of Cortisol and Melatonin. This is a configurable assay, and either or both analytes can be selected. This kit may be used for the analysis of human serum, plasma, tissue/cell lysate, culture supernatant samples, or cerebrospinal fluid (CSF). Serum or plasma samples should be extracted using either the acetonitrile precipitation technique or the Waters 96-well HLB extraction plate. Please refer to the protocol for more information on both methods. The necessary components to perform the extractions are not included with this kit.

Panel Type: Neuroscience

Specificity

Cross Reactivty
There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes within a panel.

Application

  • Analytes: Cortisol, Melatonin
  • Recommended Sample type: serum, plasma, cerebrospinal fluid (CSF), tissue/cell lysate, and culture supernatant samples
  • NOTE: Sample extraction protocols are required for serum/plasma samples. Other sample types (e.g. CSF, culture supernatant, saliva, etc.) do not require sample extraction.
  • Recommended Sample dilution: 1:4 for extracted serum and plasma or neat for CSF and culture supernatant
  • Assay Run Time: Overnight
  • Research Category: Neuroscience Metabolism
  • Research Subcategory: Category Metabolic Disorders Obesity Inflammation & Autoimmune Mechanisms
  • NOTE: This is a competitive assay

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

96-well plate
Everything you need in a single kit.

Components

Human Circadian/Stress Standard, 1 vial, lyophilized

Human Circadian/Stress Quality Control 1, 1 vial, lyophilized

Human Circadian/Stress Quality Control 2, 1 vial, lyophilized

Assay Buffer, 2 bottles, 30 mL ea

Wash Buffer, 10X (0.05% Proclin), 2 bottles, 30 mL ea

Human Circadian/Stress Primary Antibodies, 1 bottle, 3.5 mL

Human Circadian/Stress Detection Antibodies, 1 bottle, 5.5 mL

Human Circadian/Stress Antibody-Immobilized Magnetic Beads, 20X, 200 uL ea

Streptavidin-Phycoerythrin, 1 bottle, 5.5 mL

Mixing Bottle, 1 bottle

Sample Plate, 96 well format, 1 plate with 2 Foil Plate Sealers

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mi Ran Choi et al.
Journal of behavioral addictions, 9(1), 93-104 (2020-01-21)
Overindulgence in Internet gaming, which is related to rapid development of the online game industry, can cause a psychiatric disorder known as Internet gaming disorder (IGD). The number of adolescents with IGD is on the rise in countries with developed

Related Content

Learn how multiplex detection of Alzheimer’s disease biomarkers, using the MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel, allows for a different perspective on neurodegenerative disease research.

Learn how multiplex detection of Alzheimer’s disease biomarkers, using the MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel, allows for a different perspective on neurodegenerative disease research.

Learn how multiplex detection of Alzheimer’s disease biomarkers, using the MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel, allows for a different perspective on neurodegenerative disease research.

Learn how multiplex detection of Alzheimer’s disease biomarkers, using the MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel, allows for a different perspective on neurodegenerative disease research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service