HCYTA-60K-PXBK48
MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A 48 Plex Premixed Magnetic Bead Panel – BULK (Space Saver) PACKAGING - Immunology Multiplex Assay
Synonym(s):
Human cytokine multiplex kit, Luminex® human cytokine immunoassay, Millipore human cytokine panel
About This Item
Recommended Products
species reactivity
human
Quality Level
manufacturer/tradename
Milliplex®
technique(s)
multiplexing: suitable
detection method
fluorometric (Luminex® xMAP®)
storage temp.
2-8°C
1 of 4
This Item | MABS2195 | MABS2194 | AV44276 |
---|---|---|---|
human ... CYBB(1536) | human ... CYBB(1536) | human ... CYBB(1536) | human ... CYBB(1536) |
human | human | human | rabbit, mouse, human, rat |
unconjugated | unconjugated | unconjugated | unconjugated |
mouse | mouse | mouse | - |
NL7, monoclonal | 54.1, monoclonal | CL5, monoclonal | polyclonal |
General description
Application
Features and Benefits
Packaging
Storage and Stability
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2
Target Organs
Respiratory Tract
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
See how a MILLIPLEX® multiplex cytokine assay demonstrated superior lot-to-lot consistency of cytokine measurement in human serum and plasma samples when compared to other Luminex® technology-based multiplex assays.
See how a MILLIPLEX® multiplex cytokine assay demonstrated superior lot-to-lot consistency of cytokine measurement in human serum and plasma samples when compared to other Luminex® technology-based multiplex assays.
See how MILLIPLEX® multiplex cytokine panels can be used in alternate sample types such as human tears, urine, milk, saliva, and CSF with an example of characterizing neuroinflammation across healthy, MCI, and AD CSF samples.
See how MILLIPLEX® multiplex cytokine panels can be used in alternate sample types such as human tears, urine, milk, saliva, and CSF with an example of characterizing neuroinflammation across healthy, MCI, and AD CSF samples.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service