Skip to Content
Merck
All Photos(1)

Key Documents

HATF00010

Millipore

Immobilon® -NC Nitrocellulose Membrane

1 roll, 33 cm x 3 m, 0.45 µm pore size, Triton-free, nitrocellulose-based transfer membrane

Synonym(s):

Western blotting membrane, blotting membrane, nitrocellulose transfer membrane, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.22

Product Name

Immobilon®-NC Transfer Membrane, 1 roll, 33 cm x 3 m, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane

material

mixed cellulose esters (MCE) membrane
nitrocellulose membrane
plain filter
white filter

Quality Level

feature

hydrophilic

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

filter L × W

33 cm × 3 m

pore size

0.45 μm pore size

capacity

117 μg/cm2 adsorption capacity (insulin)
160 μg/cm2 adsorption capacity (BSA)
259 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with India ink
for use with Ponceau-S red

detection method

chemiluminescent
colorimetric
fluorometric
radioactive

shipped in

ambient

General description

Immobilon-NC (HATF) membrane is composed of a mixed cellulose esters matrix. It is devoid of surfactants that may interfere with cell wall integrity during cell growth. This membrane is recommended for colony and plaque lifts.

Application

Immobilon-NC Transfer Membrane has been used in western blot analysis.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunhua Hou et al.
Experimental cell research, 387(2), 111750-111750 (2019-12-02)
Lymphoma, a malignant tumor, is mainly characterized by painless lymph node enlargement and hepatosplenomegaly. At present, lymphoma is mainly treated by radiation, chemical drugs, bone marrow transplantation and surgery. However, due to the high degree of heterogeneity, lymphomas are highly
Bing Wang et al.
Oncology letters, 13(5), 3494-3500 (2017-05-23)
The aim of the present study was to gain insight into the molecular mechanism of gefitinib resistance in non-small cell lung cancer (NSCLC), and demonstrate whether long noncoding RNA (lncRNA) expression signatures differ between gefitinib-sensitive PC9 and gefitinib-resistant PC9 (PC9-R)
Kanika Verma et al.
Nature communications, 11(1), 2926-2926 (2020-06-12)
Metabolic changes alter the cellular milieu; can this also change intracellular protein folding? Since proteostasis can modulate mutational buffering, if change in metabolism has the ability to change protein folding, arguably, it should also alter mutational buffering. Here we find
1H-Indole-3-Carbonyl-Thiazole-4-Carboxylic Acid Methyl Ester Blocked Human Glioma Cell Invasion via Aryl Hydrocarbon Receptor?s Regulation of Cytoskeletal Contraction
Zhao L, et al.
BioMed Research International (2020)
Lin Liu et al.
OncoTargets and therapy, 12, 11507-11516 (2020-01-11)
ARHGAP10 belongs to the ARHGAP family, which is downregulated in certain human tumors. However, the detailed function of ARHGAP10 remains unclear in human colon carcinoma (CRC). In the current study, we aimed to explore the role of ARHGAP10 in the

Protocols

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service