Skip to Content
Merck
All Photos(1)

Key Documents

FC010

Sigma-Aldrich

Human Fibronectin

from human plasma, liquid, 1 mg/mL, purified protein, suitable for cell culture

Synonym(s):

Fibronectin protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75
Pricing and availability is not currently available.

Product Name

Human Plasma Fibronectin Purified Protein, from human plasma, liquid, 1 mg/mL (100 MG pack size is lyophilized), purified protein, suitable for cell culture

biological source

human

Quality Level

Assay

~95% (SDS-PAGE)

form

(liquid: 1MG, 5MG, 10MG pack size
lyophilized: 100 MG pack size)

mol wt

220 kDa

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

cell culture | mammalian: suitable

surface coverage

2—10 μg/cm2

input

sample type mesenchymal stem cell(s)
sample type neural stem cell(s)
sample type: human embryonic stem cell(s)
sample type epithelial cells
sample type hematopoietic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type pancreatic stem cell(s)

UniProt accession no.

Binding Specificity

Peptide Source: Collagen

Peptide Source: Laminin

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... FN1(2335)

General description

Fibronectins (FNs) are high-molecular-mass adhesive glycoproteins of the extracellular matrix (ECM) and body fluids. It is a dimer composed of two subunits linked by two disulfide bonds. Each monomer of FN consists of three types of repeating units: 12 FN repeats of type I (approximately 40 amino acids each), two type II repeats (around 60 amino acids), and 15-17 type III repeats (approximately 90 amino acids). These repeating units contribute to the structural and functional diversity of fibronectin molecules.[1]

Application

Human fibronectin (HFN) is suitable for use as an attachment factor in the propagation of cells in vitro when used to coat cell culture surfaces, including plastic ware, glassware, and microcarrier beads.[2]

Biochem/physiol Actions

Fibronectin plays a crucial role in mediating diverse cellular interactions and is involved in various processes, including cell adhesion, the establishment and maintenance of cell morphology, cell migration, growth, and differentiation. It interacts with several extracellular matrix (ECM) and cell surface proteins, such as collagen, heparin, fibrin, and specific cell membrane receptors. Fibronectin can also serve as a ligand for multiple integrins, including the well-known fibronectin receptor alpha5-beta1, which binds to the arginylglycylaspartic acid (RGD) sequence in the repeat III-10 region of fibronectin. This extensive network of interactions highlights fibronectin′s significance in regulating cellular behavior and mediating tissue function.[1]

Physical form

Liquid in 50 mM tris-buffered saline (TBS), pH 7.5,containing no preservatives.Lyophilized from 50 mM tris-buffered saline (TBS), pH 7.5,containing no preservatives (100 MG pack size). Reconstitute the entire vial ofhFN to 1 mg/mL using a sterile balanced salt solution at 37°C.

Storage and Stability

Maintain at 2-8°C for up to 6 months from date of receipt. Do not freeze.

Analysis Note

A double band of 220 kDa is present under reduced conditions.hFN is purified by affinity chromatography on gelatin agarose, followed by chromatography on heparin-agarose.Plasma from donors has been screened, and shown to be negative for HIV, HTLV, Hepatitis B and C.

Other Notes

Suggested Procedure for Coating Cell Cultureware

1.Determine the amount of HFN needed to coat culture vessels by multiplying the total surface area (cm2) by the desired concentration (μg/mL) of HFN. Recommended amount is 2-10 μg/cm2.

2.Wet the surface of each culture vessel to be coated with a minimum amount of sterile balanced salt solution (serum and protein free) required to cover the entire area.

3.Introduce the proper CO2 atmosphere, if required.

4.Add the calculated amount of HFN to each culture vessel.

5.Allow HFN to adsorb to the surface of the vessel for 5-20 minutes.

6.Remove residual balanced salt solution before proceeding with standard cell culture procedures


Product is filtered using a 0.1 micron filter during production. It is recommended to filter product again before use/after reconstitution.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alternative splicing of the angiogenesis associated extra-domain B of fibronectin regulates the accessibility of the B-C loop of the type III repeat 8
Elisa Ventura, et al.
PLoS ONE, 5(2), e9145-e9145 (2010)
Aleksander A Rebane et al.
FEBS letters, 594(7), 1132-1144 (2019-12-14)
Golgins are an abundant class of peripheral membrane proteins of the Golgi. These very long (50-400 nm) rod-like proteins initially capture cognate transport vesicles, thus enabling subsequent SNARE-mediated membrane fusion. Here, we explore the hypothesis that in addition to serving as
Sandrine B Lavenus et al.
The Journal of biological chemistry, 295(19), 6700-6709 (2020-04-03)
Tumor cells can spread to distant sites through their ability to switch between mesenchymal and amoeboid (bleb-based) migration. Because of this difference, inhibitors of metastasis must account for each migration mode. However, the role of vimentin in amoeboid migration has
Lan Huang et al.
Bio-protocol, 10(2), e3487-e3487 (2020-01-20)
Infantile hemangioma (IH) is a vascular tumor noted for its excessive blood vessel formation during infancy, glucose-transporter-1 (GLUT1)-positive staining of the blood vessels, and its slow spontaneous involution over several years in early childhood. For most children, IH poses no
Teemu Suutari et al.
Analytical chemistry, 92(21), 14509-14516 (2020-10-16)
Real-time label-free techniques are used to profile G protein-coupled receptor (GPCR) signaling pathways in living cells. However, interpreting the label-free signal responses is challenging, and previously reported methods do not reliably separate pathways from each other. In this study, a

Articles

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

See All

Protocols

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Questions

  1. What are other close alternatives for cell culture applications?

    What are other close alternatives for cell culture applications?

    1 answer
    1. A number of options for fibronectin derived from human plasma are available for cell culture applications. Please refer to our pre-faceted product table for Attachment Factors:

      https://www.sigmaaldrich.com/products/cell-culture-and-analysis/cell-culture-supplements-and-reagents/attachment-factors?facets=facet_biological_source%3Ahuman&term=fibronectin

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service