EZSH0USIN1
EZ-Stream® Pump Housing Kit
for use with EZ-Stream® Vacuum Pumps
Select a Size
Vendor SKU: AA10-2071-250MG
A$996.15
Select a Size
About This Item
Vendor SKU: AA10-2071-250MG
A$996.15
Recommended Products
feature
repair kit
manufacturer/tradename
EZ-Stream®
application(s)
bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring
compatibility
for use with EZ-Stream® Vacuum Pumps
General description
The EZ-Stream® Vacuum Pump is ideal to be used both on work bench and laminar flow hood, where its small footprint saves laboratory space.
Application
Components
- Front panel.
- Back panel.
- Base with ID sticker.
- Horseshoe.
- Assembley Screws.
Legal Information
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service