Skip to Content
Merck
All Photos(1)

Key Documents

EZMSAA3

Millipore

Mouse SAA-3 ELISA Kit

measures and quantifies SAA-3 levels in 10 μL serum or plasma

Synonym(s):

Serum amyloid A-3 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

Product Name

Mouse SAA-3 ELISA, This Mouse SAA-3 ELISA is used to measure & quantify SAA-3 levels in Metabolism & Cancer & Neuroscience research.

Quality Level

species reactivity

mouse

packaging

kit of 1 × 96 wells

parameter

10 μL sample volume (4hr assay)

assay range

accuracy: 96-100%
sensitivity: 0.078 μg/mL
(10 μl sample size)

standard curve range: 0.078-5.0 μg/mL

technique(s)

ELISA: suitable

input

sample type cell culture supernatant
sample type plasma (K2 EDTA)
sample type serum

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

General description

Millipore is pleased to announce the release of the Mouse SAA-3 ELISA kit. This kit will analyze up to 38 samples in duplicate, requiring as little as 10 ul of mouse serum, plasma or tissue culture supernatants. The availability of a mouse SAA-3 ELISA kit should facilitate additional research studies with regards to body weight regulation and metabolism, cardiovascular disease and immune regulation.
Serum Amyloid A3 (SAA-3) is an acute phase protein produced in the liver. In response to injury, granulocytes and macrophages secrete cytokines which modulate the production of acute phase proteins. Many of these proteins, such as SAA-3, α1-acid glycoprotein (AGP), lipoclain 24P3, plasminogen activator inhibitor-1 (PAI-1) and PTX3 are secreted from adipocytes and are elevated in various metabolic disease states.

Application

Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Metabolic Disorders

Obesity
This assay requires 10 µl of undiluted sample and is performed at room temperature as a 4 hour assay.
Used to detect/quantify: SAA-3

Storage and Stability

Prior to use, all components in the kit can be stored up to 2 weeks at 2-8°C.

Other Notes

Please contact Technical Service for linearity of dilution.

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service