Skip to Content
Merck
All Photos(1)

Key Documents

CBL582

Sigma-Aldrich

Anti-CD19 Antibody, clone HD37

clone HD37, Chemicon®, from mouse

Synonym(s):

B-lymphocyte Surface Antigen B4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

HD37, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... CD19(930)

Specificity

The CD19 antigen (90 kDa) is expressed from the earliest stages of B-progenitor development, on all peripheral B cells including germinal centre B cells and all B cell lines and B cell leukaemias tested. T cell and monocytic cell lines are negative and the antigen is lost on B cell maturations to plasma cells. The antigen is a type I integral membrane glycoprotein whose in vitro inhibition will influence B cell activation and proliferation.

Application

Anti-CD19 Antibody, clone HD37 detects level of CD19 & has been published & validated for use in FC, IP, IH.
Identification of CD19 positive B lymphocytes and malignant B cells by indirect immunofluorescence staining

Leukaemia and lymphoma phenotyping

Identification of normal and malignant CD19 positive B lymphocytes in cryostat frozen sections

Immunoprecipitation of the CD19 antigen

Studies of the CD19/21 complex in B cell activation and the control of B cell development

Optimal working dilutions must be determined by the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Linkage

Replaces: CBL1322

Physical form

Format: Purified
The monoclonal is presented at a concentration of 100μg/1ml in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Storage and Stability

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylvia Eash et al.
The American journal of pathology, 164(2), 419-428 (2004-01-27)
JC virus (JCV), a member of the polyomavirus family, causes a demyelinating disease of the central nervous system (CNS) in humans known as progressive multifocal leukoencephalopathy. Although glial cells are the principal target of JCV productive infection in progressive multifocal
G F Fischer et al.
Journal of immunology (Baltimore, Md. : 1950), 144(2), 638-641 (1990-01-15)
The CD24 Ag is present on human B cells from the earliest stages of B lineage development and is lost on terminal B cell maturation to plasma cells. This Ag is also expressed on mature forms of granulocytes. We studied
A Greiner et al.
Developmental immunology, 6(3-4), 187-195 (1998-11-14)
Low-grade MALT-type lymphomas are malignancies of mucosal marginal-zone B cells and preceded by reactive inflammatory lymphoid tissue. Experimental observations suggest that antigen and CD40 Ligand act during cognate T/B cell interaction and are crucial for germinal center B-cell maturation generating
Gladys S Ingle et al.
British journal of haematology, 140(1), 46-58 (2007-11-10)
CD19 and CD21 (CR2) are co-receptors found on B-cells and various B-cell lymphomas, including non-Hodgkin lymphoma. To evaluate their suitability as targets for therapy of such lymphomas using internalization-dependent antibody-drug conjugates [such as antibody-4-(N-maleimidomethyl)cyclohexane-1-carboxylate, (N2'-deacetyl-N2'-(3-mercapto-1-oxopropyl)-maytansine) (MCC-DM1) conjugates, which require lysosomal
Maristella Maggi et al.
Cancers, 13(22) (2021-11-28)
E. coli L-asparaginase is an amidohydrolase (EC 3.5.1.1) which has been successfully used for the treatment of Acute Lymphoblastic Leukemia for over 50 years. Despite its efficacy, its side effects, and especially its intrinsic immunogenicity, hamper its usage in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service