Skip to Content
Merck
All Photos(1)

Key Documents

CBL553

Sigma-Aldrich

Anti-CD63 Antibody, clone RFAC4

clone RFAC4, Chemicon®, from mouse

Synonym(s):

Anti-LAMP-3, Anti-ME491, Anti-MLA1, Anti-OMA81H, Anti-TSPAN30

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

RFAC4, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD63(967)

Specificity

This antibody reacts with a 53 kDa protein which form a part of the family of tetraspan moieties. The antigen was originally designated as a lysosomal membrane protein characterized as an activation dependent platelet surface antigen. In fact the CD63 antigen has a diverse distribution on the surface and in the cytoplasm of many cell types including lymphoid, myeloid, endothelial cells and melanoma.

Application

Anti-CD63 Antibody, clone RFAC4 is an antibody against CD63 for use in FC, IP, WB, IH, IH(P).
Flow cytometric studies of CD63 expression

Immunoprecipitation

Western blotting of CD63 antigen from solubilized platelet proteins

Suitable for formalin fixed paraffin wax embedded tissue sections. HIER and proteolytic pretreatment may not be necessary. Users should confirm specificity and pretreatment for their particular application.

Optimal working dilutions must be determined by the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Physical form

Format: Purified
The monoclonal is presented at a concentration of 100μg/1ml in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Storage and Stability

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mariëtte E G Kranendonk et al.
Obesity (Silver Spring, Md.), 22(5), 1296-1308 (2013-12-18)
Extracellular vesicles (EVs) released by human adipocytes or adipose tissue (AT)-explants play a role in the paracrine interaction between adipocytes and macrophages, a key mechanism in AT inflammation, leading to metabolic complications like insulin resistance (IR) were determined. EVs released
Haruka Shinohara et al.
Journal of immunology (Baltimore, Md. : 1950), 199(4), 1505-1515 (2017-07-12)
Macrophages are polarized into functional classically activated and alternatively activated (M2) phenotypes depending on their microenvironment, and these cells play an important role in the immune system. M2-like polarization of tumor-associated macrophages (TAMs) is activated by various secretions from cancer
Yonatan Perez et al.
Brain : a journal of neurology, 140(4), 928-939 (2017-03-24)
A novel autosomal recessive cerebro-renal syndrome was identified in consanguineous Bedouin kindred: neurological deterioration was evident as of early age, progressing into severe intellectual disability, profound ataxia, camptocormia and oculomotor apraxia. Brain MRI was normal. Four of the six affected
Valentina Salvi et al.
JCI insight, 3(10) (2018-05-18)
The excessive production of type I IFNs is a hallmark and a main pathogenic mechanism of many autoimmune diseases, including systemic lupus erythematosus (SLE). In these pathologies, the sustained secretion of type I IFNs is dependent on the improper activation
Bing Xia et al.
Theranostics, 10(20), 8974-8995 (2020-08-18)
Rationale: Peripheral nerves are unique in their remarkable elasticity. Schwann cells (SCs), important components of the peripheral nervous system (PNS), are constantly subjected to physiological and mechanical stresses from dynamic stretching and compression forces during movement. So far, it is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service