Skip to Content
Merck
All Photos(1)

Key Documents

AP187C

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, Cy3 conjugate, Species Adsorbed

1.5 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

CY3 conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

concentration

1.5 mg/mL

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Rabbit IgG (H+L) (absorbed for minimal cross-reactivity to Human, Mouse and Rat Serum Proteins)

FLUOROPHORE/ PROTEIN ABSORBANCE RATIO

Approximately 2.25-2.8(A552/A280) = 2.7

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling
Suggested dilution for most applications: 1:100-1:800 for most applications.

Optimal working dilutions must be determined by the end user.
This Goat anti-Rabbit IgG Antibody, Cy3 conjugate, Species Adsorbed is validated for use in IF for the detection of Rabbit IgG.

Physical form

Lyophilized. Buffer = 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 7.6 with 15 mg/mL BSA, and 0.05 Sodium Azide.

RECONSTITUTION:

Reconstitute to 1.5 mg/mL with sterile distilled water.

Storage and Stability

Maintain lyophilized product at 2-8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kang Chen et al.
Molecular medicine reports, 13(3), 2439-2446 (2016-02-06)
Pseudomonas aeruginosa (PA) is a common Gram‑negative bacterium and can cause serious infections, including hospital‑acquired pneumonia, suppurative bacterial keratitis and acute burn wound infection. The pathogenesis of PA infections is closely associated with excessive inflammatory responses and bacterial virulence factors.
Xufeng Dai et al.
PloS one, 12(11), e0188032-e0188032 (2017-11-14)
Adeno-associated virus (AAV) vectors are important gene delivery tools for the treatment of many recessively inherited retinal diseases. For example, a wild-type (WT) AAV5 vector can deliver a full-length Cnga3 (cyclic nucleotide-gated channel alpha-3) cDNA to target cells of the
Estrogen and androgen receptor expression in surface epithelium and inclusion cyst in the ovary of premenopausal and postmenopausal women.
Mendez, C; Morales-Vasquez, F; Perez-Montiel, D; Gomora, MJ; Espinola-Zetina et al.
Journal of Ovarian Research null
Kayoko Hosaka et al.
Cancer communications (London, England), 43(6), 637-660 (2023-04-30)
Tumors possess incessant growth features, and expansion of their masses demands sufficient oxygen supply by red blood cells (RBCs). In adult mammals, the bone marrow (BM) is the main organ regulating hematopoiesis with dedicated manners. Other than BM, extramedullary hematopoiesis
Xufeng Dai et al.
PloS one, 11(5), e0156542-e0156542 (2016-05-27)
Lysophosphatidylcholine acyltransferase 1 (LPCAT1) is necessary for photoreceptors to generate an important lipid component of their membranes. The absence of LPCAT1 results in early and rapid rod and cone degeneration. Retinal degeneration 11 (rd11) mice carry a mutation in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service