AGX806
2019-nCoV Nucleocapsid Recombinant Antigen,full length (Ag-6)
recombinant, expressed in E. coli, suitable for lateral flow assay
Synonym(s):
SARS-CoV, SARS-CoV-2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.26
Recommended Products
recombinant
expressed in E. coli
Quality Level
form
liquid
technique(s)
immunodiffusion: suitable
lateral flow assay: suitable
UniProt accession no.
shipped in
dry ice
Gene Information
SARS coronavirus ... N(43740575)
General description
This E. coli recombinant protein 2019-nCoV Nucleocapsid Recombinant Antigen,full length (Ag-6), Cat. No. AGX806 is tested for use in Lateral Flow Assay.
Specificity
SARS-CoV-2
Target description
Coronavirus disease 2019 (COVID-19) is an infectious disease caused by the virus strain severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), a virus closely related to the SARS virus. A SARS-CoV-2 nucleocapsid antigen can be used to support the detection of SARS-CoV-2 (COVID-19, 2019-nCoV).
Physical form
Purified
Column Purification
Purified protein in 10 mM Phosphate buffer, 150 mM NaCl with no preservatives, pH 7.4.
Column Purification
Purified protein in 10 mM Phosphate buffer, 150 mM NaCl with no preservatives, pH 7.4.
Storage and Stability
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service