Skip to Content
Merck
All Photos(1)

Key Documents

ACS5030

Millipore

Amicon® Pro Purification System with Amicon® Ultra-0.5 Device

Amicon Pro Purification System - A 30 kDa Centrifugal Tool collapsing protein purifcation with concentraion & buffer exchange.

Synonym(s):

Amicon Ultra-0.5 System, Pro Purification System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24

material

Amicon Ultra styrene-butadiene device
Ultracel® regenerated cellulose membrane (Amicon Ultra-0.5; low binding)
polyethylene cap
polyethylene frit
polypropylene device (for exchange)
polypropylene frit
polypropylene tube (AU0.5; for collection)
polypropylene tube (for collection)
polypropylene tube (holder)

sterility

non-sterile

product line

Amicon®

manufacturer/tradename

Amicon® Pro

parameter

10 mL sample volume

technique(s)

buffer exchange: suitable
protein purification: suitable (antibody labeling)
protein purification: suitable (concentration)
protein purification: suitable (enrichment)

L

12 cm (4.7 in.)

diam.

2.97 cm

filtration area

1 cm2

pore size

30 kDa NMWCO

shipped in

ambient

General description

Amicon® Pro Purification System device streamlines the affinity purification process by providing a single adaptable centrifugation unit capable of performing all steps in the affinity purification process. Attaching the Amicon 0.5 mL filter during the elution step provides simultaneous concentration and the option of highly efficient diafiltration (99%) in a single spin.

Application

Amicon Pro Purification System - A 30 kDa Centrifugal Tool. Ideal for collapsing protein purification with concentration & buffer exchange, protein enrichment, and antibody labeling.
Amicon® Pro Purification System with Amicon Ultra-0.5 Device is suitable for affinity purification, depleting abundant proteins from serum samples, and desalting from chromatography samples.

Features and Benefits

  • It can tackle the most liable and poorly expressed proteins
  • Simple protein sample preparation
  • Reduced risk during sample transfer and hands-on time
  • Time-saving and enables protein recovery
  • Higher protein retention and specific activity

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Filtration methods and membrane filter selection in nanoparticle purification and production.

Filtration methods and membrane filter selection in nanoparticle purification and production.

Filtration methods and membrane filter selection in nanoparticle purification and production.

Filtration methods and membrane filter selection in nanoparticle purification and production.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service