Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
mouse
concentration
0.5 mg/mL
technique(s)
immunohistochemistry: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
mouse ... Postn(50706)
General description
Immunogen
Application
Cell Structure
ECM Proteins
Immunohistochemistry Analysis: A representative lot detected Periostin (Pan) in certain reported tissues (Kruzynska-Frejtag, A., et al. (2004). Dev Dyn. 229(4):857-868.; Butcher, J. T., et al. (2007). Dev Biol. 302(1):256-266.; Norris, R. A., et al. (2007). J Cell Biochem. 101(3):695-711.; Inai, K., et al. (2008). Dev Biol. 315(2):383-396.; Norris, R. A., et al. (2008). Dev Biol. 316(2):200-213.).
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 1 µg/mL of this antibody detected Periostin (Pan) in 10 µg of mouse lung tissue lysate.
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source rabbit | biological source rabbit | biological source mouse | biological source mouse |
| shipped in wet ice | shipped in wet ice | shipped in ambient | shipped in dry ice |
| species reactivity mouse | species reactivity rat, mouse, human | species reactivity human | species reactivity human |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
wgk
WGK 2
Storage Class
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



