Skip to Content
Merck
All Photos(2)

Key Documents

ABS2185

Sigma-Aldrich

Anti-Methyl-Glutamine

from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG

shipped in

ambient

General description

Glutamine methylation occurs on translation termination factors and ribosomal proteins. Protein methylation has been reported to affect various cellular processes including protein stability, protein-protein interaction, and protein localization. Methylation of histone is suggested as a platform to recruit and regulate other chromatin-related factors. Glutamine methylation is brought about by the action of protein glutamine methyltransferase HEMK2 that specifically requires a GQX3R motif for methylation activity. This antibody was tested for glutamine methylation in chromodomain helicase DNA-binding protein 5 (CHD5) that contains multiple PHD and chromo domains and is reported to function as a tumor suppressor.

Specificity

This rabbit polyclonal antibody detects methylglutamine in proteins.

Immunogen

KLH-conjugated Gly-Gln (Me)-Gly tripeptide.

Application

Anti-Methyl-Glutamine, Cat. No. ABS2185, is a rabbit polyclonal antibody that detects glutamine methylated proteins. It has been tested for use in Western Blotting.
Western Blotting Analysis: A 1:3,000 dilution from a representative lot detected Methyl-Glutamine in CHD5 unmethylated & CHD5 methylated recombinant lysate (Coutesy of, Dr. Kusevic, Dr. Kuditipudi and Dr. Jeltsch at University Stuttgart, Germany).

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected Methyl-Glutamine in in-vitro methylated CHD5, NUT, and eRF1 proteins (Kusevic, D., et. al. (2016). J Biol Chem. 291(12):6124-33).

Quality

Evaluated by Western Blotting in methylated recombinant chromodomain helicase DNA-binding 5 (CHD5) protein, but not in unmethylated recombinant CHD5.

Western Blotting Analysis: A 1:3,000 dilution of this antibody detected Methyl-Glutamine in methylated recombinant chromodomain helicase DNA-binding 5 (CHD5) protein, but not in unmethylated recombinant CHD5.

Target description

~51 kDa observed (for methylated CHD5). Uncharacterized bands may be observed in some lysate(s).

Physical form

Antiserum was depleted by running over unmethylated protein column.
Format: Unpurified
Rabbit polyclonal antiserum with 0.05% sodium azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service