Skip to Content
Merck
All Photos(1)

Key Documents

ABN297

Sigma-Aldrich

Anti-Oxytocin receptor Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Oxytocin receptor, OT-R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
mouse, human
technique(s):
western blot: suitable
citations:

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... OXTR(5021)

General description

Oxytocin is a cyclic 9 amino acid peptide that differs from vasopressin in 2 amino acids. Despite the close similarities in sequence, oxytocin and vasopressin have different biological activities and bind to distinct G protein-coupled receptors. The oxytocin receptor, OT, couples primarily to Gq/11 to mobilize intracellular calcium. In female reproduction, oxytocin promotes uterine contraction and lactation; oxytocin is the most commonly used drug for induction of labor, whereas an oxytocin antagonist, atosiban, is under investigation to suppress preterm labor. Oxytocin/OT interaction in the CNS also plays an important role in stress, male and female sexual response, and sociality.

Immunogen

Epitope: Cytoplasmic domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human Oxytocin receptor.

Application

Anti-Oxytocin receptor Antibody is a highly specific rabbit polyclonal antibody, that targets Oxytocin Receptor & has been tested in western blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in MCF7 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Oxytocin receptor in 10 µg of MCF7 cell lysate.

Target description

~45/65 kDa observed The calculated molecular weight of this protein is 43 kDa However, this protein has been observed at ~60-70 kDa due to glycoslyatation (Zhong, M.m et al. (2012). Molecular Cancer Research. 1-25). Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service