ABE1298
Anti-phospho BAF60C (Ser247)
from rabbit, purified by affinity chromatography
Synonym(s):
SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 3, 60 kDa BRG-1/Brm-associated factor subunit C, BRG1-associated factor 60C, BAF60C
About This Item
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
mouse
species reactivity (predicted by homology)
human (based on 100% sequence homology), rat (based on 100% sequence homology), bovine (based on 100% sequence homology)
packaging
antibody small pack of 25 μL
technique(s)
inhibition assay: suitable (peptide)
western blot: suitable
NCBI accession no.
UniProt accession no.
target post-translational modification
phosphorylation (pSer247)
Gene Information
human ... SMARCD3(6604)
General description
Specificity
Immunogen
Application
Quality
Western Blotting Analysis: A 1:500 dilution of this antibody detected phospho BAF60C (Ser247) in NIH3T3 Nuclear Extract.
Target description
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service