Skip to Content
Merck
All Photos(2)

Key Documents

ABC929

Sigma-Aldrich

Anti-MAP1LC3A antibody

rabbit polyclonal

Synonym(s):

Microtubule-associated proteins 1A/1B light chain 3A, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, MAP1 light chain 3-like protein 1, MAP1A/MAP1B light chain 3 A, Microtubule-associated protein 1 light chain 3 alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-LC3-I/II Antibody, from rabbit, purified by affinity chromatography

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

Xenopus (based on 100% sequence homology), rat (based on 100% sequence homology), porcine (based on 100% sequence homology), mouse (based on 100% sequence homology), zebrafish (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Microtubule-associated LC3A constitute nearly half of the mass of all the microtubule associated proteins that copurify with brain microtubules. LC3A is composed of a heavy chain and multiple light-chain subunits. The molecular machinery required for autophagy is highly conserved in all eukaryotes as seen by the high degree of conservation of proteins involved in the formation of the autophagosome membranes. Recently, both yeast Apg8p and its rat homologue LC3-II were identified as essential constituents of autophagosome membrane as a processed form. In addition, both the yeast and human proteins exist in two modified forms produced by a series of post-translational modifications including a critical C-terminal cleavage after a conserved Gly residue, and the smaller processed form is associated with the autophagosome membranes Results revealed different regulation of the three human isoforms of LC3-II and implicate that the three isoforms may have different physiological functions. Two forms of LC3A, called LC3-I and -II, were produced post-translationally in various cells. LC3-I is cytosolic, whereas LC3-II is membrane bound. LC3-II is the first mammalian protein identified that specifically associates with autophagosome membranes.

Specificity

This antibody detects both the non-lipidated LC3-I (non-autophagic) and lipidated LC3-II (autophagic) under autophagy conditions.

Immunogen

Recombinant protein corresponding to Human LC3-I/II.

Application

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected LC3 in serum-starved, chloroquine-treated HeLa cells. A punctate distribution is seen, evidence of autophagy conditions within the cell.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-LC3-I/II Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of LC3-I/II.

Quality

Evaluated by Western Blotting in serum-starved, chloroquine-treated HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected LC3-I (non-lipidated) and LC3-II (lipidated) in 10 µg of serum-starved, chloroquine-treated HeLa cell lysate.

Target description

~17/14 kDa observed

Linkage

Replaces: MABC177; MABC176

Physical form

Antigen Affinity Purified
Purified rabbit Polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Angel Merlos Rodrigo et al.
Cells, 8(3) (2019-03-16)
The efficiency of cisplatin (CDDP) is significantly hindered by the development of resistance during the treatment course. To gain a detailed understanding of the molecular mechanisms underlying the development of cisplatin resistance, we comparatively analyzed established a CDDP-resistant neuroblastoma cell
Chen Wang et al.
Cell death & disease, 12(11), 1055-1055 (2021-11-08)
Cervical cancer is the leading cause of cancer-related deaths in women, and treatment for cervical cancer is very limited. Emerging evidence suggests that targeting ferroptosis is a promising way to treat cancer. Here, we investigated the role of ferroptosis in
Francesco Petragnano et al.
International journal of radiation biology, 96(6), 823-835 (2020-03-10)
Purpose: Radiation therapy (RT), by using ionizing radiation (IR), destroys cancer cells inducing DNA damage. Despite several studies are continuously performed to identify the best curative dose of IR, the role of dose-rate, IR delivered per unit of time, on
Jin Wang et al.
International journal of oncology, 58(1), 70-82 (2020-12-29)
W922, a novel PI3K/Akt/mTOR pathway inhibitor, exhibits efficient anti‑tumor effects on HCT116, MCF‑7 and A549 human cancer cells compared with other synthesized compounds. The present study aimed to investigate its anti‑tumor effects on colorectal cancer cells. A total, of seven
Pengzhi Chen et al.
Experimental and therapeutic medicine, 21(5), 440-440 (2021-03-23)
In the pathogenesis of diabetic cataract, high glucose levels induce oxidative damage in human lens epithelial cells (HLECs). Resveratrol has been demonstrated to be a potent antioxidant in various disease conditions; however, limited information is available on its effects on

Articles

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service