Skip to Content
Merck
All Photos(2)

Key Documents

ABC348

Sigma-Aldrich

Anti-Bmf Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Bcl-2-modifying factor, Bmf

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
WB
species reactivity:
human
technique(s):
immunohistochemistry: suitable
western blot: suitable
citations:

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BMF(90427)

General description

Bcl-2-modifying factor (Bmf) is a member of the Bcl-2 family containing one BCL2 homology domain 3. Members of this family are critical regulators of apoptosis by either inhibiting or promoting cell death. Bmf associates with the dynein light chain 2 (DLC2) component of the myosin V motors and is sequestered by the cell′s actin cytoskeleton. Bmf is constitutively expressed in many tissues. There are 3 isoforms of Bmf. Isoform 1 is mainly expressed in B-lymphoid cells while isoform 2 and 3 are mainly expressed in B-CLL and normal B-cells.

Specificity

This antibody recognizes the N-terminus of Bmf.

Immunogen

KLH-conjugated linear peptide corresponding to the N-terminus of human Bmf.

Application

Detect Bcl-2-modifying factor using this rabbit polyclonal antibody, Anti-Bmf Antibody validated for use in western blotting & IHC.
Immunocytochemistry Analysis: 10 µg/mL from a representative lot detected Bmf in HeLa cells.

Quality

Evaluated by Western Blotting in HepG2 and HEK293 cell lysates.

Western Blotting Analysis: 1 µg/mL of this antibody detected Bmf in 15 µg of HepG2 and HEK293 cell lysates.

Target description

~25 kDa observed

Analysis Note

Control
HepG2 and HEK293 cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service