Skip to Content
Merck
All Photos(1)

Key Documents

AB9848

Sigma-Aldrich

Anti-phospho-Synapsin I (Ser62,Ser67) Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer62/pSer67)

Gene Information

human ... SYN1(6853)

Specificity

Synapsin I, phospho Serine 62,67. The antibody recognizes a protein doublet of ~78 kDa corresponding to Synapsin I, phospho Serine 62,67 in lysates from rat cortex. Immunolableing is blocked by preadsorption with the phospho-peptide used as the immunogen but not by the corresponding dephospho-peptide.
The immunogen has 100% homology with mouse and bovine.

Immunogen

Synthetic peptide of amino acids surrounding the phosphoSerine 62,67 site of rat Synapsin I.

Application

Anti-phospho-Synapsin I Antibody (Ser62, Ser67) detects level of phospho-Synapsin I (Ser62) & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology

Physical form

ImmunoAffinity Purified
Liquid in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug/mL BSA and 50% glycerol.

Storage and Stability

Maintain at -20degC in undiluted for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles. Do not store in a self defrosting freezer.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chongbo Zhong et al.
Frontiers in neural circuits, 16, 978837-978837 (2022-10-11)
Modulation of the release of glutamate by activation of presynaptic nicotinic acetylcholine receptors (nAChRs) is one of the most prevalent mechanism of nicotinic facilitation of glutamatergic transmission in cortico-limbic circuits. By imaging gene chimeric co-cultures from mouse, we examined the
Soledad Bárez-López et al.
Molecular & cellular proteomics : MCP, 22(5), 100544-100544 (2023-04-09)
The cell bodies of hypothalamic magnocellular neurones are densely packed in the hypothalamic supraoptic nucleus, whereas their axons project to the anatomically discrete posterior pituitary gland. We have taken advantage of this unique anatomical structure to establish proteome and phosphoproteome
Soledad Bárez-López et al.
Neuroendocrinology, 113(10), 1008-1023 (2023-06-05)
Despite the widespread use of general anaesthetics, the mechanisms mediating their effects are still not understood. Although suppressed in most parts of the brain, neuronal activity, as measured by FOS activation, is increased in the hypothalamic supraoptic nucleus (SON) by
Julie Fourneau et al.
Journal of neurochemistry, 147(2), 240-255 (2018-05-29)
In human, a chronic sensorimotor perturbation (SMP) through prolonged body immobilization alters motor task performance through a combination of peripheral and central factors. Studies performed on a rat model of SMP have shown biomolecular changes and a reorganization of sensorimotor
Ellen R Bowen et al.
Discover mental health, 3(1), 27-27 (2023-12-01)
Schizophrenia is a debilitating condition necessitating more efficacious therapies. Previous studies suggested that schizophrenia development is associated with aberrant synaptic pruning by glial cells. We pursued an interdisciplinary approach to understand whether therapeutic reduction in glial cell-specifically astrocytic-phagocytosis might benefit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service