Skip to Content
Merck
All Photos(1)

Key Documents

AB9678

Sigma-Aldrich

Anti-GABA A Receptor α 5 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GABRA5(2558)

Specificity

GABAA Receptor alpha 5 subunit. The antibody recognizes a protein of ~55 kDa corresponding to GABAA Receptor alpha 5 subunit in lysates from rat hippocampus and thalamus.

Immunogen

Fusion protein from the Cytosolic loop of rat the GABAA Receptor alpha 5 subunit.

Application

Anti-GABA A Receptor α 5 Antibody is an antibody against GABA A Receptor α 5 for use in WB.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Li et al.
Frontiers in neurology, 12, 591223-591223 (2021-03-13)
Post-operative cognitive dysfunction (POCD) is a common complication during the post-operative period. It affects the recovery time of the patient after surgery and the stay time in hospital, which causes a great deal of burden to patients and families emotionally
Kohei Koga et al.
Molecular pain, 14, 1744806918783478-1744806918783478 (2018-06-30)
Background Chronic pain is a persistent unpleasant sensation that produces pathological synaptic plasticity in the central nervous system. Both human imaging study and animal studies consistently demonstrate that the anterior cingulate cortex is a critical cortical area for nociceptive and
Guobin Xia et al.
Molecular psychiatry, 26(7), 2837-2853 (2021-03-27)
The high comorbidity between obesity and mental disorders, such as depression and anxiety, often exacerbates metabolic and neurological symptoms significantly. However, neural mechanisms that underlie reciprocal control of feeding and mental states are largely elusive. Here we report that melanocortin
George Gallos et al.
American journal of physiology. Lung cellular and molecular physiology, 308(9), L931-L942 (2015-02-11)
The clinical need for novel bronchodilators for the treatment of bronchoconstrictive diseases remains a major medical issue. Modulation of airway smooth muscle (ASM) chloride via GABAA receptor activation to achieve relaxation of precontracted ASM represents a potentially beneficial therapeutic option.
Muhammad Hassan et al.
International journal of molecular sciences, 23(24) (2022-12-24)
Absence seizures are hyperexcitations within the cortico-thalamocortical (CTC) network, however the underlying causative mechanisms at the cellular and molecular level are still being elucidated and appear to be multifactorial. Dysfunctional feed-forward inhibition (FFI) is implicated as one cause of absence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service