Skip to Content
Merck
All Photos(1)

Key Documents

AB5847

Sigma-Aldrich

Anti-Glutamate Receptor 1 Antibody, phosphoSer 831

Chemicon®, from rabbit

Synonym(s):

GluR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

packaging

antibody small pack of 25 μL

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIA1(2890)

Specificity

Recognizes Glutamate Receptor 1 [GluR1], phosphoSer831. The antibody recognizes a protein of ~100 kDa corresponding to GluR1 phosphorylated at Ser831 in rat hippocampal homogenate. Immunolabeling of the GluR1 protein is blocked by the immunogen peptide but not by the corresponding non-phosphopeptide. The immunogen sequence is 100% homologous in human and mouse.

Immunogen

Epitope: phosphoSer 831
Synthetic peptide corresponding to amino acids surrounding the phosphoSer831 of rat GluR1.

Application

Detect Glutamate Receptor 1 using this Anti-Glutamate Receptor 1 Antibody, phosphoSer 831 validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot and dot blots: 1:500-1:1000. The working dilution should be based on the tissue and the expected phosphorylation state.

Optimal working dilutions must be determined by the end user.

Target description

100 kDa

Linkage

Replaces: 05-823

Physical form

Affinity Purified immunoglobulin. Liquid in 10 mM HEPES, pH 7.5, with 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.
ImmunoAffinity Purified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Hippocampal neurons, rat brain tissue lysates or mouse brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumasri Guntupalli et al.
The Journal of biological chemistry, 292(20), 8186-8194 (2017-04-06)
The accumulation of soluble amyloid-β (Aβ) peptides produces profound neuronal changes in the brain during the pathogenesis of Alzheimer's disease. Excessive levels of Aβ disrupt excitatory synaptic transmission by promoting the removal of synaptic AMPA receptors (AMPARs), dendritic spine loss
Mónica Fernández-Monreal et al.
Journal of cell science, 129(14), 2793-2803 (2016-06-04)
Hippocampal synaptic plasticity involves both membrane trafficking events and intracellular signaling, but how these are coordinated is far from clear. The endosomal transport of glutamate receptors in and out of the postsynaptic membrane responds to multiple signaling cascades triggered by
Marion Rame et al.
PloS one, 12(5), e0177036-e0177036 (2017-05-05)
Single sub-anesthetic doses of ketamine can exacerbate the symptoms of patients diagnosed with schizophrenia, yet similar ketamine treatments rapidly reduce depressive symptoms in major depression. Acute doses of the atypical antipsychotic drug clozapine have also been shown to counteract ketamine-induced
Jocelyn Widagdo et al.
Cellular and molecular neurobiology, 40(7), 1213-1222 (2020-02-14)
Excitatory neurotransmission relies on the precise targeting of α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA)-type glutamate receptors to the neuronal plasma membrane. Activity-dependent ubiquitination of AMPA receptor (AMPAR) subunits sorts internalised receptors to late endosomes for degradation, which ultimately determines the number of AMPARs
Men C Tan et al.
Frontiers in cellular neuroscience, 11, 124-124 (2017-05-19)
Transcription of new RNA is crucial for maintaining synaptic plasticity, learning and memory. Although the importance of synaptic plasticity-related messenger RNAs (mRNAs) is well established, the role of a large group of long non-coding RNAs (lncRNAs) in long-term potentiation (LTP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service