Skip to Content
Merck
All Photos(1)

Key Documents

AB5832

Sigma-Aldrich

Anti-BACE Antibody

Chemicon®, from rabbit

Synonym(s):

ASP2, BACE1, Beta Secretase, beta-Site APP Cleaving Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BACE1(23621)

Specificity

Reacts with human BACE by Western blot.

Immunogen

Synthetic peptide corresponding to amino acids 458 to 501 of human BACE (Vassar et al. 1999).

Application

Anti-BACE Antibody is an antibody against BACE for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western blotting: 0.5-1.0 μg/mL on human brain tissue. The antibody recognizes a protein of molecular weight of 70 kDa in normal human brain lysate.

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid. Buffer = PBS containing 0.02% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Tanokashira et al.
eNeuro, 2(2) (2015-10-16)
The β-secretase called BACE1 is a membrane-associated protease that initiates the generation of amyloid β-protein (Aβ), a key event in Alzheimer's disease (AD). However, the mechanism of intraneuronal regulation of BACE1 is poorly understood. Here, we present evidence that low-density
Naomi Mamada et al.
Molecular brain, 8(1), 73-73 (2015-11-11)
β-Site amyloid precursor protein cleaving enzyme 1 (BACE1) is a membrane-bound aspartyl protease that initiates amyloid β-protein (Aβ) generation. Aberrant elevation of BACE1 levels in brains of Alzheimer's disease (AD) patients may involve Aβ. In the present study, we used
Gurdeep Marwarha et al.
Journal of neurochemistry, 144(6), 761-779 (2018-01-10)
The etiology of Alzheimer's disease (AD) is egregiously comprehended, but epidemiological studies have posited that diets rich in the saturated fatty acid palmitic acid (palmitate) are a significant risk factor. The production and accumulation of amyloid beta peptide (Aβ) is
Gurdeep Marwarha et al.
Molecular neurobiology, 56(7), 5256-5269 (2018-12-21)
Numerous cross-sectional and longitudinal studies have implicated saturated fat-enriched diets in the etio-pathogenesis of Alzheimer's disease (AD). Emerging evidence shows that saturated fat-enriched diets, such as palmitate-enriched diets, increase amyloid-beta (Aβ) production, the histopathological hallmark of AD. However, the molecular
D Porquet et al.
Age (Dordrecht, Netherlands), 37(1), 9747-9747 (2015-02-11)
The amyloid precursor protein/presenilin 1 (APP/PS1) mouse model of Alzheimer's disease (AD) has provided robust neuropathological hallmarks of familial AD-like pattern at early ages, whereas senescence-accelerated mouse prone 8 (SAMP8) has a remarkable early senescence phenotype with pathological similarities to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service