Skip to Content
Merck
All Photos(1)

Documents

AB1846

Sigma-Aldrich

Anti-C-X-C Chemokine Receptor 4 Antibody, NT

Chemicon®, from rabbit

Synonym(s):

CD184, CXCR4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CXCR4(7852)

Specificity

Recognizes CXCR4, N-terminus.

Immunogen

Amino-terminus peptide of human CXCR4 (Berson, 1996; Leotscher, 1994).
Epitope: N-terminus

Application

Anti-C-X-C Chemokine Receptor 4 Antibody, N-terminus is an antibody against C-X-C Chemokine Receptor 4 for use in IC, IP & WB.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
Western blot: 1-2 μg/mL. HeLa cell lysates can be used as positive control. The antibody should detect a band at 40 kDa.

Immunocytochemistry: 10 μg/mL

Immunoprecipitation

Optimal working dilutions must be determined by end user.

Target description

40 kDa

Physical form

Format: Purified
Protein A purified
Purified IgG in PBS containing 0.02% sodium azide.

Storage and Stability

Maintain 1 year at 4°C, from date of shipment. Do not freeze. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Skin

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barani Govindarajan et al.
Scientific reports, 13(1), 20634-20634 (2023-11-24)
The chemokine receptor, CXCR4 signaling regulates cell growth, invasion, and metastasis to the bone-marrow niche in prostate cancer (PCa). Previously, we established that CXCR4 interacts with phosphatidylinositol 4-kinase IIIα (PI4KIIIα encoded by PI4KA) through its adaptor proteins and PI4KA overexpressed
Xu Cui et al.
Journal of neuroscience research, 87(1), 86-95 (2008-08-20)
We tested the hypothesis that a nitric oxide donor, DETA-NONOate, up-regulates stromal cell-derived factor-1 (SDF1) and angiopoietin 1 (Ang1) in the ischemic brain and their respective receptors chemokine CXC motif receptor 4 (CXCR4) and Tie2 in the subventricular zone (SVZ)
Linhui Wang et al.
Oncology reports, 22(6), 1333-1339 (2009-11-04)
Renal cell carcinoma (RCC) shows organ-specific metastasis. This may be attributed to the fact that the CXCR4 G protein coupled receptor on RCC cells mediates chemo-attraction toward stromal-derived factor (SDF)-1 secreted by target organs. SDF-1 binding to CXCR4 initiates many
Nitric oxide donor upregulation of stromal cell-derived factor-1/chemokine (CXC motif) receptor 4 enhances bone marrow stromal cell migration into ischemic brain after stroke.
Cui, X; Chen, J; Zacharek, A; Li, Y; Roberts, C; Kapke, A; Savant-Bhonsale, S; Chopp, M
Stem Cells null
Juan Xu et al.
PloS one, 8(3), e56114-e56114 (2013-03-09)
The latent membrane protein 1 (LMP1), which is encoded by the Epstein-Barr virus (EBV), is an important oncogenic protein that is closely related to carcinogenesis and metastasis of nasopharyngeal carcinoma (NPC), a prevalent cancer in China. We previously reported that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service