Skip to Content
Merck
All Photos(1)

Key Documents

AB1558

Sigma-Aldrich

Anti-Dopamine D2 Receptor Antibody

serum, Chemicon®

Synonym(s):

Anti-D2DR, Anti-D2R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

rat ... Drd2(24318)

Specificity

Recognizes Dopamine D2 Receptor (D2R). Recognizes both native and denatured forms of D2R (110, 68 and 47 kDa bands by Western blot using rat striatal positive control (Catalog Number AG282)). AB1558 inhibits binding of [3H]YM-09152-2, a known D2R antagonist and (125)I-NAPS, a photoligand. It inhibits binding of [3H]YM-09152-2 and PPHT-fluorescein (an agonist of D2R) and reacts with 110 kDa D2R band in fibroblasts transfected with rat D2R cDNA. AG284 is available for blocking experiments; AG284 is not recommended as a positive control for western blots.

Immunogen

Amino-terminal peptide sequence (aa 24-34) near or at the ligand binding domain of rat D2R (1,2).

The immunogen shows 100% homology with mouse, 90% with human and bovine and 72% with canine D2R receptors. An antibody made to the same sequence has shown reactivity with human, lobster, squid and crawfish.

Application



Immunohistochemistry: 4% PFA fixed, frozen sections, 1:200, 24 hour incubation at room temperature, DAB detection. {Tillerson, JL et al. (2006) Behavioural Brain Res. 172:97-105.

Immunocytochemistry: 4% PFA fixed cells 10-15′, permeabilized with 0.2-0.3% triton X-100, 1:100, o/N 4C for fluorescence {Sun, W et al (2005) Stem Cells 23(7):931-945, http://stemcells.alphamedpress.org/cgi/content/full/23/7/931}.

Western blot: 1:1,000-1:5,000 (Chemiluminescence technique) Higher concentration of antibody may be necessary when using low concentration samples or when using milk in antibody diluents. It may be necessary to load 100-200 μg of purified striatal or other tissue membranes to visualize the DR2 bands. The antibody has been shown to recognize both native and denatured DR2 bands.

ELISA: 1:10,000-1:50,000 [1 μg/mL D2R peptide (Cat. Number AG284)/well]

Optimal working dilutions must be determined by end user.
Anti-Dopamine D2 Receptor Antibody is an antibody against Dopamine D2 Receptor for use in ELISA, IC, IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Target description

110, 68 and 47 kDa

Physical form

Rabbit antiSerum. Liquid containing 0.05% sodium azide.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Brain

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandre Morin et al.
Frontiers in behavioral neuroscience, 15, 770374-770374 (2022-01-22)
Global cerebral ischemia (GCI) in rats has been shown to promote exploration of anxiogenic zones of the Elevated-Plus Maze (EPM) and Open Field Test (OFT). This study investigated changes in impulsive choice and/or defensive responses as possible contributors of heightened
Violeta-Maria Caragea et al.
Brain structure & function, 229(3), 609-637 (2023-08-24)
The dopaminergic (DA) system regulates both motor function, and learning and memory. The cerebellum supports motor control and the acquisition of procedural memories, including goal-directed behavior, and is subjected to DA control. Its fastigial nucleus (FN) controls and interprets body
K E Borgmann-Winter et al.
Neuroscience, 158(2), 642-653 (2008-11-11)
The olfactory epithelium constitutes the sole source of regenerating neural cells that can be obtained from a living human. As such, primary cultures derived from human olfactory epithelial biopsies can be utilized to study neurobiological characteristics of individuals under different
Jickssa M Gemechu et al.
Scientific reports, 8(1), 1517-1517 (2018-01-26)
Mutations in parkin gene (Park2) are linked to early-onset autosomal recessive Parkinson's disease (PD) and young-onset sporadic PD. Park2 knockout (PKO) rodents; however, do not display neurodegeneration of the nigrostriatal pathway, suggesting age-dependent compensatory changes. Our goal was to examine
Ren-Hong Du et al.
Journal of neuroinflammation, 15(1), 258-258 (2018-09-12)
α-Synuclein (α-Syn)-induced neuroinflammation plays a crucial role in the pathogenesis of Parkinson's disease (PD). Dopamine D2 receptor (Drd2) has been regarded as a potential anti-inflammatory target in the therapy of neurodegenerative diseases. However, the effect of astrocytic Drd2 in α-Syn-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service