Skip to Content
Merck
All Photos(1)

Key Documents

AB1297

Sigma-Aldrich

Anti-Corticosterone Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

sheep

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
radioimmunoassay: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Corticosterone

Cross-reactivity: 11-dehydrocorticosterone 0.67%, Deoxycorticosterone 1.5%, 18-OH-DOC <0.01%, Cortisone <0.01%, Cortisol <0.01% and Aldosterone 0.2% by RIA.



Cross-reactivity: 11-dehydrocorticosterone 0.35%, Progesterone 0.004%, 18-OH-DOC 0.01%, Cortisol 0.12%, 18-OH-B 0.02% and Aldosterone 0.06% by ELISA.

Immunogen

Corticosterone-3-cmo-urease

Application

Anti-Corticosterone Antibody is an antibody against Corticosterone for use in ELISA, RIA.
ELISA: 1:200,000 (Sensitivity 5 pg/well, id-50 is 50 pg/well.)

RIA: 1:60,000 (Sensitivity 10 pg/tube, 50% displacement 60 pg/tube separating free and bound using ammonium sulfate 10 50% saturation.)

Optimal working dilutions must be determined by end user.
Research Category
Metabolism
Research Sub Category
Metabolic Hormones & Receptors

Physical form

Sheep serum. Liquid in 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 6 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanja Hofmann et al.
Poultry science, 100(8), 101243-101243 (2021-06-28)
Already during early life, chickens need to cope with chronic stressors that can impair their health and welfare, with stocking density being one of the most influential factors. Nevertheless, there is a gap in research on the influence of stocking
Jose Gulfo et al.
PloS one, 11(1), e0146497-e0146497 (2016-01-08)
Corticosteroid-binding globulin (CBG) is the specific plasma transport glycoprotein for glucocorticoids. Circulating CBG is mainly synthesized in liver but, its synthesis has been located also in other organs as placenta, kidney and adipose tissue with unknown role. Using an experimental
Lorraine Jaimes-Hoy et al.
Endocrinology, 157(8), 3253-3265 (2016-06-21)
Hypothalamic-pituitary-thyroid (HPT) axis activity is important for energy homeostasis, and is modified by stress. Maternal separation (MS) alters the stress response and predisposes to metabolic disturbances in the adult. We therefore studied the effect of MS on adult HPT axis
Sonja Schmucker et al.
Poultry science, 100(11), 101408-101408 (2021-09-17)
During life, the number and function of immune cells change with potential consequences for immunocompetence of an organism. In laying hens, studies have primarily focused on early development of immune competence and only few have investigated systemic and lymphatic distribution
Sex Differences in Serotonin 5-HT 1A Receptor Responses to Repeated Restraint Stress in Adult Male and Female Rats.
Philippe, et al.
The International Journal of Neuropsychopharmacology, 25, 863-876 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service