8.55045
Rink Amide MBHA resin LL (100-200 mesh)
Novabiochem®
Synonym(s):
Rink Amide MBHA resin LL (100-200 mesh), 4-(2Æ,4Æ-Dimethoxyphenyl-Fmoc-aminomethyl)-phenoxyacetamido norleucyl-MBHA resin (100-200 mesh)
About This Item
Recommended Products
Quality Level
product line
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
amine
storage temp.
2-30°C
SMILES string
N(C(c7c(cc(cc7)OC)OC)c4ccc(cc4)OCC(=O)NC(c6ccc(cc6)C)c5ccc(cc5)C)C(=O)OCC1c2c(cccc2)c3c1cccc3
InChI key
JDDWRLPTKIOUOF-UHFFFAOYSA-N
Related Categories
General description
Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Linkage
Analysis Note
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.30 - 0.40 mmol/g
TNBS-Test: passes test
Swelling Volume (in DMF): lot specific result
Swelling Volume (in CH₂Cl₂): lot specific result
Total swelling volume acc. Houben Weyl (in CH2Cl2): ≥ 5.2 ml/g
Total swelling volume acc. Houben Weyl (in DMF): ≥ 5.2 ml/g
Identity (of the substitution): passes test
The polymer matrix is copoly (styrene-1 % DVB) 100 - 200 mesh.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.
Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.
Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.
Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.
Protocols
Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.
Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.
Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.
Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service