Skip to Content
Merck
All Photos(1)

Key Documents

8.22314

Sigma-Aldrich

4-Methoxybenzaldehyde

for synthesis

Synonym(s):

4-Methoxybenzaldehyde, Anisaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€54.20
1 L
€182.00

€54.20


Estimated to ship on06 May 2025


To order products, please contact your local dealer.

Select a Size

Change View
250 ML
€54.20
1 L
€182.00

About This Item

Linear Formula:
4-(CH3O)C6H4CHO
CAS Number:
Molecular Weight:
136.15
MDL number:
UNSPSC Code:
12352114
EC Index Number:
204-602-6
NACRES:
NA.22

€54.20


Estimated to ship on06 May 2025


To order products, please contact your local dealer.

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

220 °C

potency

3200 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-5.3 % (v/v)

refractive index

n20/D 1.57

pH

7 (20 °C, 2 g/L in H2O)

bp

247-249 °C/1013 hPa

transition temp

flash point 116 °C

solubility

2 g/L

density

1.12 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

O(C)c1ccc(cc1)C=O

InChI

1S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3

InChI key

ZRSNZINYAWTAHE-UHFFFAOYSA-N

Application

<ul>
<li><strong>Multi-Target Directed Ligands for Alzheimer′s Disease: </strong>Used as a drug fragment to synthesize novel substituted thiosemicarbazones for the treatment of Alzheimer′s disease (AD) (Varma et al., 2023).</li>
<li><strong>Wound Dressing Membranes Based on Immobilized Anisaldehyde:</strong> wound dressing membranes involved immobilizing anisaldehyde (4-Methoxybenzaldehyde) onto a chitosan-GA-gelatin copolymer, offering insights into its therapeutic potential in wound care applications (Tamer et al., 2022).</li>
</ul>

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.121 - 1.124
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service