Skip to Content
Merck
All Photos(1)

Key Documents

8.22288

Sigma-Aldrich

Periodic acid

for synthesis

Synonym(s):

Periodic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H5IO6
CAS Number:
Molecular Weight:
227.94
MDL number:
UNSPSC Code:
12352301
EC Index Number:
233-937-0
NACRES:
NA.22

Quality Level

Assay

≥98% (iodometric)

form

solid

reaction suitability

reagent type: oxidant

pH

1.2 (20 °C, 100 g/L in H2O)

mp

122 °C

solubility

ethanol: soluble at 20 °C
water: soluble at 20 °C

bulk density

1400 kg/m3

storage temp.

15-25°C

SMILES string

[I](=O)(O)(O)(O)(O)O

InChI

1S/H5IO6/c2-1(3,4,5,6)7/h(H5,2,3,4,5,6,7)

InChI key

TWLXDPFBEPBAQB-UHFFFAOYSA-N

Application

Periodic acid can be used as a reagent for the oxidative cleavage of 1,2 diols to ketones or aldehydes, and epoxides to aldehydes. It is also used in the oxidative cleavage of various protecting groups such as acetonide, tetrahydropyranyl ether (THP), ethoxyethyl acetals, cyclic acetals, dithioacetals, and oxathioacetals.
It can also be used:
  • In the selective N-nitrosation of secondary amines in the presence of sodium nitrite, and wet silica gel.
  • As a stoichiometric oxidant with chromium (IV) acetate for the oxidation of tertiary C−H bonds to tertiary alcohols.
  • In combination with chromium oxide to oxidize primary alcohols to carboxylic acids; alkylbenzenes to benzoic acids; and cyclic benzyl ethers to the corresponding lactones.
  • In the iodination of bulky polyalkyl benzenes in the presence of iodine.
  • In the oxidative coupling of polycyclic aromatic hydrocarbons in the presence of aqueous acetic acid.
  • In the selective oxidation of sulfides to sulfoxides using FeCl3 as a catalyst.

Analysis Note

Assay (iodometric): ≥ 98.0 %
Melting range (lower value): ≥ 124 °C
Melting range (upper value): ≤ 129 °C

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1B - STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Periodic Acid
Stengel JH, et al.
eEROS (Encyclopedia of Reagents for Organic Synthesis), 1-6 (2001)
One-pot hydrothermal synthesis and characterization of CoFe2O4 nanoparticles and its application as magnetically recoverable catalyst in oxidation of alcohols by periodic acid
Paul B, et al.
Materials Chemistry and Physics, 181, 99-105 (2016)
A mild and highly efficient oxidation of sulfides to sulfoxides with periodic acid catalyzed by FeCl3
Kim SS, et al.
Synthesis, 2002(17), 2484-2486 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service