Skip to Content
Merck
All Photos(1)

Key Documents

8.20851

Sigma-Aldrich

2,7-Naphthalenediol

for synthesis

Synonym(s):

2,7-Naphthalenediol, 2,7-Dihydroxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O2
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352104
EC Index Number:
209-478-7
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

powder

mp

184-187 °C

bulk density

600‑700 kg/m3

storage temp.

2-30°C

SMILES string

Oc1cc2c(cc1)ccc(c2)O

InChI

1S/C10H8O2/c11-9-3-1-7-2-4-10(12)6-8(7)5-9/h1-6,11-12H

InChI key

DFQICHCWIIJABH-UHFFFAOYSA-N

Application

  • Photoelectric Properties of Carbon Quantum Dots: Investigates the synthesis and high fluorescent quantum yield of green carbon quantum dots, where 2,7-naphthalenediol serves as a precursor, enhancing the photoelectric properties for potential applications in optoelectronic devices (Zheng et al., 2020).
  • Fungal Melanogenesis Mimicry for Coatings: Describes the use of dihydroxynaphthalene, derived from 2,7-naphthalenediol, in creating multifunctional coatings that mimic fungal melanogenesis, showcasing its application in protecting materials and increasing their durability (Jeon et al., 2016).
  • Catalytic and SERS Activities of Nanoclusters: Discusses the role of 2,7-naphthalenediol in enhancing the catalytic and Surface-Enhanced Raman Scattering (SERS) activities of osmium nanoclusters, which are crucial for chemical sensing and catalytic applications (Ede et al., 2014).
  • Shape-Selective Pt Nanoparticles: Details the photoinduced formation of shape-selective platinum nanoparticles, where 2,7-naphthalenediol could potentially act as a reducing agent, influencing the synthesis process in photoreactive nanomaterial production (Kundu and Liang, 2010).
  • Glycolate Testing in Microbial Cells: Presents a new rapid glycolate testing method using diethyl ether extraction suitable for small bacterial cell samples. While primarily a methodological study, it highlights the broader potential applications of reagents like 2,7-naphthalenediol in biochemical testing (Uchida et al., 1999).

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service