Skip to Content
Merck
All Photos(1)

Key Documents

8.18682

Sigma-Aldrich

Thiophene-2-ethanol

for synthesis

Synonym(s):

Thiophene-2-ethanol, 2-(2-Thienyl)ethanol, 2-(2-Hydroxyethyl)thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8OS
CAS Number:
Molecular Weight:
128.19
MDL number:
UNSPSC Code:
12352104
EC Index Number:
226-452-0
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

liquid

transition temp

flash point 105 °C

density

1.15 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[s]1c(ccc1)CCO

InChI

1S/C6H8OS/c7-4-3-6-2-1-5-8-6/h1-2,5,7H,3-4H2

InChI key

VMJOFTHFJMLIKL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.150 - 1.160
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service